missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylcyclopentane-1,3-dione, 98%, Thermo Scientific Chemicals

$65.37 - $997.10

Chemical Identifiers

CAS 765-69-5
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001406
InChI Key HXZILEQYFQYQCE-UHFFFAOYSA-N
Synonym 2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46
PubChem CID 13005
IUPAC Name 2-methylcyclopentane-1,3-dione
SMILES CC1C(=O)CCC1=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1393606
View Documents
Thermo Scientific Chemicals
A1393606
5 g N/A
 
AAA1393614
View Documents
Thermo Scientific Chemicals
A1393614
25 g N/A
 
AAA1393622
View Documents
Thermo Scientific Chemicals
A1393622
100 g N/A
 
Description

Description

2-Methylcyclopentane-1,3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone. The alkylation of 2-methylcyclopentane-1,3-dione (A) with 2-(1-naphthyl)ethyl bromide gives only the O-alkylation product. Mixtures of C- and O-alkyl derivatives are obtained. Michael addition of 2-methylcyclopentane-1,3-dione to methyl vinyl ketone in water gives 2-methyl-2-(3-oxo- butyl)-1,3-cyclopentanedione (3), an important intermediate of natural product chemistry. 2-Methylcyclopentane-1, 3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclopentane-1,3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone. The alkylation of 2-methylcyclopentane-1,3-dione (A) with 2-(1-naphthyl)ethyl bromide gives only the O-alkylation product. Mixtures of C- and O-alkyl derivatives are obtained. Michael addition of 2-methylcyclopentane-1,3-dione to methyl vinyl ketone in water gives 2-methyl-2-(3-oxo- butyl)-1,3-cyclopentanedione (3), an important intermediate of natural product chemistry. 2-Methylcyclopentane-1, 3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone.

Solubility
Solubility in hot Methanol (almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

765-69-5
112.128
HXZILEQYFQYQCE-UHFFFAOYSA-N
13005
CC1C(=O)CCC1=O
C6H8O2
MFCD00001406
2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46
2-methylcyclopentane-1,3-dione

Specifications

765-69-5
C6H8O2
5 g
2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46
CC1C(=O)CCC1=O
112.128
112.13
2-Methylcyclopentane-1,3-dione
212°C to 216°C
MFCD00001406
1237255
HXZILEQYFQYQCE-UHFFFAOYSA-N
2-methylcyclopentane-1,3-dione
13005
98%
Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 212-153-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only