missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylcyclopentanone, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 247660250

Catalog No. AC247660250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Methylcyclopentanone
98.5
0.9100g/mL
26°C
99%
C6H10O
MFCD00001414
0.91
Solubility in water: soluble. Other solubilities: very soluble in ethanol,ether and acetone
CC1CCCC1=O
98.15
98.14
1120-72-5
100.0
139°C to 140°C
Authentic
Glass bottle
1.4340 to 1.436
25g
2-methylcyclopentanone, cyclopentanone, 2-methyl, .alpha.-methylcyclopentanone, cyclopentanone, methyl, alpha-methylcyclopentanone, cyclopentan-1-one, 2-methyl, methylcyclopentanone, 2-methyl cyclopentanone, 3-methyl-2-cyclopentanone, 2-methyl-cyclopentan-1-one
ZIXLDMFVRPABBX-UHFFFAOYNA-N
2-methylcyclopentan-1-one
14265
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-318-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only