Learn More
2-Methylcyclopentanone, 98%, Thermo Scientific Chemicals
$249.88 - $249.88
Chemical Identifiers
CAS | 1120-72-5 |
---|---|
Molecular Formula | C6H10O |
Molecular Weight (g/mol) | 98.15 |
MDL Number | MFCD00001414 |
InChI Key | ZIXLDMFVRPABBX-UHFFFAOYNA-N |
Synonym | 2-methylcyclopentanone, cyclopentanone, 2-methyl, .alpha.-methylcyclopentanone, cyclopentanone, methyl, alpha-methylcyclopentanone, cyclopentan-1-one, 2-methyl, methylcyclopentanone, 2-methyl cyclopentanone, 3-methyl-2-cyclopentanone, 2-methyl-cyclopentan-1-one |
PubChem CID | 14265 |
IUPAC Name | 2-methylcyclopentan-1-one |
SMILES | CC1CCCC1=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC247660050
|
Thermo Scientific Chemicals
247660050 |
5 g | Glass bottle |
Each for $249.88
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1120-72-5 | |
98.15 | |
ZIXLDMFVRPABBX-UHFFFAOYNA-N | |
14265 | |
CC1CCCC1=O |
C6H10O | |
MFCD00001414 | |
2-methylcyclopentanone, cyclopentanone, 2-methyl, .alpha.-methylcyclopentanone, cyclopentanone, methyl, alpha-methylcyclopentanone, cyclopentan-1-one, 2-methyl, methylcyclopentanone, 2-methyl cyclopentanone, 3-methyl-2-cyclopentanone, 2-methyl-cyclopentan-1-one | |
2-methylcyclopentan-1-one |
Specifications
1120-72-5 | |
100.0 | |
0.9100g/mL | |
26°C | |
99% | |
C6H10O | |
MFCD00001414 | |
0.91 | |
Solubility in water: soluble. Other solubilities: very soluble in ethanol,ether and acetone | |
CC1CCCC1=O | |
98.15 | |
98.14 | |
Liquid |
98.5 | |
Colorless to Yellow | |
139°C to 140°C | |
Authentic | |
Glass bottle | |
1.4340 to 1.436 | |
5 g | |
2-methylcyclopentanone, cyclopentanone, 2-methyl, .alpha.-methylcyclopentanone, cyclopentanone, methyl, alpha-methylcyclopentanone, cyclopentan-1-one, 2-methyl, methylcyclopentanone, 2-methyl cyclopentanone, 3-methyl-2-cyclopentanone, 2-methyl-cyclopentan-1-one | |
ZIXLDMFVRPABBX-UHFFFAOYNA-N | |
2-methylcyclopentan-1-one | |
14265 | |
99% | |
2-Methylcyclopentanone |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 214-318-4
RUO – Research Use Only