missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylindole-3-acetic acid, 98+%, Thermo Scientific Chemicals

$54.96 - $182.25

Chemical Identifiers

CAS 1912-43-2
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00075006
InChI Key QJNNHJVSQUUHHE-UHFFFAOYSA-N
Synonym 2-methylindole-3-acetic acid, 2-methyl-1h-indol-3-yl acetic acid, 2-2-methyl-1h-indol-3-yl acetic acid, 2-methylindole-3-aceticacid, 1h-indole-3-acetic acid, 2-methyl, 2-methyl-3-indoleacetic acid, 2-2-methylindol-3-yl acetic acid, pubchem7229, acmc-209euw, 2-methylindole-3acetic acid
PubChem CID 589107
IUPAC Name 2-(2-methyl-1H-indol-3-yl)acetic acid
SMILES CC1=C(CC(O)=O)C2=CC=CC=C2N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1907103
View Documents
Thermo Scientific Chemicals
L1907103
1 g
Each for $54.96
 
AAL1907106
View Documents
Thermo Scientific Chemicals
L1907106
5 g
Each for $182.25
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1912-43-2
189.21
QJNNHJVSQUUHHE-UHFFFAOYSA-N
589107
CC1=C(CC(O)=O)C2=CC=CC=C2N1
C11H11NO2
MFCD00075006
2-methylindole-3-acetic acid, 2-methyl-1h-indol-3-yl acetic acid, 2-2-methyl-1h-indol-3-yl acetic acid, 2-methylindole-3-aceticacid, 1h-indole-3-acetic acid, 2-methyl, 2-methyl-3-indoleacetic acid, 2-2-methylindol-3-yl acetic acid, pubchem7229, acmc-209euw, 2-methylindole-3acetic acid
2-(2-methyl-1H-indol-3-yl)acetic acid

Specifications

1912-43-2
C11H11NO2
1 g
2-methylindole-3-acetic acid, 2-methyl-1h-indol-3-yl acetic acid, 2-2-methyl-1h-indol-3-yl acetic acid, 2-methylindole-3-aceticacid, 1h-indole-3-acetic acid, 2-methyl, 2-methyl-3-indoleacetic acid, 2-2-methylindol-3-yl acetic acid, pubchem7229, acmc-209euw, 2-methylindole-3acetic acid
CC1=C(CC(O)=O)C2=CC=CC=C2N1
189.21
189.22
2-Methylindole-3-acetic acid
198°C to 204°C (decomposition)
MFCD00075006
152701
QJNNHJVSQUUHHE-UHFFFAOYSA-N
2-(2-methyl-1H-indol-3-yl)acetic acid
589107
≥98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only