missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
2-Methylpiperazine, 98%, Thermo Scientific Chemicals
$198.00 - $198.00
Identifiants chimiques
CAS | 109-07-9 |
---|---|
Molecular Formula | C5H12N2 |
Molecular Weight (g/mol) | 100.16 |
InChI Key | JOMNTHCQHJPVAZ-UHFFFAOYSA-N |
Synonym | piperazine, 2-methyl, 2-methyl-piperazine, +/--2-methylpiperazine, 2methylpiperazine, 2-methylpiperazin, 2-methypiperazine, 2-methylpiperizine, 3-methylpiperazine, 2-methyl piperazine, pubchem8560 |
PubChem CID | 66057 |
IUPAC Name | 2-methylpiperazine |
SMILES | CC1CNCCN1 |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | |||||
---|---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | |||||
AC127471000
|
Thermo Scientific Chemicals
127471000 |
100 g | Glass bottle |
chaque for $198.00
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
109-07-9 | |
100.16 | |
piperazine, 2-methyl, 2-methyl-piperazine, +/--2-methylpiperazine, 2methylpiperazine, 2-methylpiperazin, 2-methypiperazine, 2-methylpiperizine, 3-methylpiperazine, 2-methyl piperazine, pubchem8560 | |
2-methylpiperazine |
C5H12N2 | |
JOMNTHCQHJPVAZ-UHFFFAOYSA-N | |
66057 | |
CC1CNCCN1 |
Spécifications
109-07-9 | |
White to Yellow | |
63°C | |
97.5% min. (GC) | |
C5H12N2 | |
23,17 | |
Solubility in water: 78% (25°C). | |
CC1CNCCN1 | |
100.16 | |
100.16 | |
Crystalline Powder, Crystals or Chunks |
61.0°C to 66.0°C | |
154.0°C to 156.0°C | |
Authentic | |
Glass bottle | |
100 g | |
piperazine, 2-methyl, 2-methyl-piperazine, +/--2-methylpiperazine, 2methylpiperazine, 2-methylpiperazin, 2-methypiperazine, 2-methylpiperizine, 3-methylpiperazine, 2-methyl piperazine, pubchem8560 | |
JOMNTHCQHJPVAZ-UHFFFAOYSA-N | |
2-methylpiperazine | |
66057 | |
98% | |
2-Methylpiperazine |
Sécurité et manipulation
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-644-2
RUO – Research Use Only