missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methylpiperidine 98.0+%, TCI America™
$33.57 - $242.92
Chemical Identifiers
CAS | 109-05-7 |
---|---|
Molecular Formula | C6H13N |
Molecular Weight (g/mol) | 99.18 |
MDL Number | MFCD00005982 |
InChI Key | NNWUEBIEOFQMSS-UHFFFAOYNA-N |
Synonym | 2-pipecoline, pipicoline, alpha-pipecoline, piperidine, 2-methyl, alpha-pipecolin, pipecoline, alpha, 2-methyl-piperidine, alpha-methylpiperidine, .alpha.-pipecolin, .alpha.-pipecoline |
PubChem CID | 7974 |
IUPAC Name | 2-methylpiperidine |
SMILES | CC1CCCCN1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
P044325ML
|
TCI America
P044325ML |
25 mL |
Each for $33.57
|
|
|||||
P0443100ML
|
TCI America
P0443100ML |
100 mL |
Each for $79.00
|
|
|||||
P0443500ML
|
TCI America
P0443500ML |
500 mL |
Each for $242.92
|
|
|||||
Chemical Identifiers
109-05-7 | |
99.18 | |
NNWUEBIEOFQMSS-UHFFFAOYNA-N | |
7974 | |
CC1CCCCN1 |
C6H13N | |
MFCD00005982 | |
2-pipecoline, pipicoline, alpha-pipecoline, piperidine, 2-methyl, alpha-pipecolin, pipecoline, alpha, 2-methyl-piperidine, alpha-methylpiperidine, .alpha.-pipecolin, .alpha.-pipecoline | |
2-methylpiperidine |
Specifications
109-05-7 | |
118°C | |
MFCD00005982 | |
1993 | |
NNWUEBIEOFQMSS-UHFFFAOYNA-N | |
2-methylpiperidine | |
7974 | |
≥98.0% (GC) | |
2-Methylpiperidine |
Yellow | |
C6H13N | |
25 mL | |
2-pipecoline, pipicoline, alpha-pipecoline, piperidine, 2-methyl, alpha-pipecolin, pipecoline, alpha, 2-methyl-piperidine, alpha-methylpiperidine, .alpha.-pipecolin, .alpha.-pipecoline | |
CC1CCCCN1 | |
99.18 | |
99.18 | |
Liquid |
Safety and Handling
EINECSNumber : (5)-0768
RTECSNumber : TN1240000
TSCA : Yes