missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Methylpropyl)boronic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 359950250

 View more versions of this product

Catalog No. AC359950250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

(2-Methylpropyl)boronic acid
97.5
Authentic
Glass bottle
MFCD00134156
04,III,1965
Solubility in water: soluble. Other solubilities: soluble in methanol
B(CC(C)C)(O)O
101.94
101.94
84110-40-7
100.0
98%
C4H11BO2
25g
isobutylboronic acid, isobutaneboronic acid, 2-methylpropyl boronic acid, 1-isobutaneboronic acid, 2-methylpropaneboronic acid, 2-methylprop-1-ylboronic acid, unii-khx76h7jhz, khx76h7jhz, 2-methyl-1-propylboronic acid, 2-methyl-1-propaneboronic acid
ZAZPDOYUCVFPOI-UHFFFAOYSA-N
2-methylpropylboronic acid
2734395
98%

Chemical Identifiers

84110-40-7
101.94
ZAZPDOYUCVFPOI-UHFFFAOYSA-N
2734395
B(CC(C)C)(O)O
C4H11BO2
MFCD00134156
isobutylboronic acid, isobutaneboronic acid, 2-methylpropyl boronic acid, 1-isobutaneboronic acid, 2-methylpropaneboronic acid, 2-methylprop-1-ylboronic acid, unii-khx76h7jhz, khx76h7jhz, 2-methyl-1-propylboronic acid, 2-methyl-1-propaneboronic acid
2-methylpropylboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only