missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyridine N-Oxide 99.0+%, TCI America™

$63.38 - $460.43

Chemical Identifiers

CAS 931-19-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00006199
InChI Key CFZKDDTWZYUZKS-UHFFFAOYSA-N
Synonym 2-picoline-n-oxide, 2-methylpyridine 1-oxide, 2-methylpyridine n-oxide, picoline n-oxide, 2-picoline n-oxide, methylpyridine 1-oxide, alpha-picoline n-oxide, pyridine, 2-methyl-, 1-oxide, 2-picoline, 1-oxide, picoline 1-oxide
PubChem CID 13602
ChEBI CHEBI:35578
IUPAC Name 2-methyl-1-oxidopyridin-1-ium
SMILES CC1=CC=CC=[N+]1[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P041825G
View Documents
TCI America
P041825G
25 g
Each for $63.38
Only null left
 
P0418500G
View Documents
TCI America
P0418500G
500 g
Each for $460.43
Only null left
 
Description

Description

Specifications

Chemical Identifiers

931-19-1
109.128
CFZKDDTWZYUZKS-UHFFFAOYSA-N
13602
2-methyl-1-oxidopyridin-1-ium
C6H7NO
MFCD00006199
2-picoline-n-oxide, 2-methylpyridine 1-oxide, 2-methylpyridine n-oxide, picoline n-oxide, 2-picoline n-oxide, methylpyridine 1-oxide, alpha-picoline n-oxide, pyridine, 2-methyl-, 1-oxide, 2-picoline, 1-oxide, picoline 1-oxide
CHEBI:35578
CC1=CC=CC=[N+]1[O-]

Specifications

931-19-1
261°C
MFCD00006199
2-picoline-n-oxide, 2-methylpyridine 1-oxide, 2-methylpyridine n-oxide, picoline n-oxide, 2-picoline n-oxide, methylpyridine 1-oxide, alpha-picoline n-oxide, pyridine, 2-methyl-, 1-oxide, 2-picoline, 1-oxide, picoline 1-oxide
CC1=CC=CC=[N+]1[O-]
109.128
CHEBI:35578
≥99.0% (GC)
2-Methylpyridine N-Oxide
White-Yellow
C6H7NO
25 g
CFZKDDTWZYUZKS-UHFFFAOYSA-N
2-methyl-1-oxidopyridin-1-ium
13602
109.13
Crystalline Lumps
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'