missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylresorcinol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 364740250

 View more versions of this product

Catalog No. AC364740250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methylresorcinol
608-25-3
100.0
White
Authentic
Glass bottle
CH3C6H3(OH)2
25 g
2-methylresorcinol, 2,6-dihydroxytoluene, 1,3-benzenediol, 2-methyl, 2-methylresorcin, resorcinol, 2-methyl, toluene-2,6-diol, 2-methyl-1,3-benzenediol, 1,3-dihydroxy-2-methylbenzene, 1,3-benzenediol, methyl, 2-methyl resorcinol
CC1=C(C=CC=C1O)O
124.14
124.14
Glassy Irregular Shaped Granules
98%
97.5
118°C to 124°C
264°C
97.5% min. (GC)
C7H8O2
MFCD00002271
06, 878
ZTMADXFOCUXMJE-UHFFFAOYSA-N
2-methylbenzene-1,3-diol
11843
98%

Chemical Identifiers

608-25-3
124.14
ZTMADXFOCUXMJE-UHFFFAOYSA-N
11843
CC1=C(C=CC=C1O)O
C7H8O2
MFCD00002271
2-methylresorcinol, 2,6-dihydroxytoluene, 1,3-benzenediol, 2-methyl, 2-methylresorcin, resorcinol, 2-methyl, toluene-2,6-diol, 2-methyl-1,3-benzenediol, 1,3-dihydroxy-2-methylbenzene, 1,3-benzenediol, methyl, 2-methyl resorcinol
2-methylbenzene-1,3-diol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-155-8

Recommended Storage : product darkens on standing

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only