missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Methylsulfonyl)ethanol, 97%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals L0315906

Catalog No. AAL0315906


Explore available promotions

Description

Description

2-(Methylsulfonyl)ethanol may be used as reagent in the synthesis of phenols from aryl fluorides. It may be used in the preparation of 2-(methylsulfonyl)ethyl N,N-bis(2-chloroethyl)phosphorodiamidate. 2-Hydroxyethyl Methyl Sulfone, is the versatile building block used in the synthesis of more complex sulfur containing compounds. It can be used for the studies of inhibitor and substrate moieties to acetylcholinesterase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylsulfonyl)ethanol may be used as reagent in the synthesis of phenols from aryl fluorides. It may be used in the preparation of 2-(methylsulfonyl)ethyl N,N-bis(2-chloroethyl)phosphorodiamidate. 2-Hydroxyethyl Methyl Sulfone,
Specifications

Specifications

2-(Methylsulfonyl)ethanol
27°C to 30°C
>110°C (230°F)
1.479
5 g
2-methylsulfonyl ethanol, 2-hydroxyethyl methyl sulfone, 2-methanesulfonylethanol, 2-methanesulfonylethan-1-ol, 2-methanesulfonyl-ethanol, 2-methanesulfonyl ethanol, ethanol, 2-methylsulfonyl, unii-5fi8eb2k0l, 2-methylsulphonyl ethanol, 2-methylsulfonyl ethyl alcohol
KFTYFTKODBWKOU-UHFFFAOYSA-N
2-methylsulfonylethanol
84834
97%
15205-66-0
148°C to 149°C (1 mmHg)
C3H8O3S
MFCD00014747
1746915
Soluble in water.
CS(=O)(=O)CCO
124.154
124.16

Chemical Identifiers

15205-66-0
124.154
KFTYFTKODBWKOU-UHFFFAOYSA-N
84834
CS(=O)(=O)CCO
C3H8O3S
MFCD00014747
2-methylsulfonyl ethanol, 2-hydroxyethyl methyl sulfone, 2-methanesulfonylethanol, 2-methanesulfonylethan-1-ol, 2-methanesulfonyl-ethanol, 2-methanesulfonyl ethanol, ethanol, 2-methylsulfonyl, unii-5fi8eb2k0l, 2-methylsulphonyl ethanol, 2-methylsulfonyl ethyl alcohol
2-methylsulfonylethanol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 239-261-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only