missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Methylsulfonylamino)benzeneboronic acid, 95%, Thermo Scientific™
$137.29 - $137.29
Chemical Identifiers
CAS | 756520-78-2 |
---|---|
Molecular Formula | C7H10BNO4S |
Molecular Weight (g/mol) | 215.03 |
MDL Number | MFCD02179471 |
InChI Key | RPRMOEFOTPSWBO-UHFFFAOYSA-N |
Synonym | 2-methylsulfonamido phenyl boronic acid, 2-methanesulfonylaminophenylboronic acid, 2-methylsulfonylaminophenylboronic acid, 2-methanesulfonamidophenyl boronic acid, 2-methanesulfonylamino phenylboronic acid, 2-methanesulfonamido phenyl boronic acid, 2-methylsulphonylamino benzeneboronic acid, 2-methanesulfonamidophenylboronic acid, 2-methylsulfonylamino benzeneboronic acid, pubchem6370 |
PubChem CID | 2773533 |
IUPAC Name | [2-(methanesulfonamido)phenyl]boronic acid |
SMILES | B(C1=CC=CC=C1NS(=O)(=O)C)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH53288MD
|
Thermo Scientific Chemicals
H53288MD |
250 mg |
Each for $137.29
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
756520-78-2 | |
215.03 | |
RPRMOEFOTPSWBO-UHFFFAOYSA-N | |
2773533 | |
B(C1=CC=CC=C1NS(=O)(=O)C)(O)O |
C7H10BNO4S | |
MFCD02179471 | |
2-methylsulfonamido phenyl boronic acid, 2-methanesulfonylaminophenylboronic acid, 2-methylsulfonylaminophenylboronic acid, 2-methanesulfonamidophenyl boronic acid, 2-methanesulfonylamino phenylboronic acid, 2-methanesulfonamido phenyl boronic acid, 2-methylsulphonylamino benzeneboronic acid, 2-methanesulfonamidophenylboronic acid, 2-methylsulfonylamino benzeneboronic acid, pubchem6370 | |
[2-(methanesulfonamido)phenyl]boronic acid |
Specifications
756520-78-2 | |
C7H10BNO4S | |
250 mg | |
RPRMOEFOTPSWBO-UHFFFAOYSA-N | |
[2-(methanesulfonamido)phenyl]boronic acid | |
2773533 | |
95% |
116°C to 120°C | |
MFCD02179471 | |
2-methylsulfonamido phenyl boronic acid, 2-methanesulfonylaminophenylboronic acid, 2-methylsulfonylaminophenylboronic acid, 2-methanesulfonamidophenyl boronic acid, 2-methanesulfonylamino phenylboronic acid, 2-methanesulfonamido phenyl boronic acid, 2-methylsulphonylamino benzeneboronic acid, 2-methanesulfonamidophenylboronic acid, 2-methylsulfonylamino benzeneboronic acid, pubchem6370 | |
B(C1=CC=CC=C1NS(=O)(=O)C)(O)O | |
215.03 | |
215.03 | |
2-(Methylsulfonylamino)benzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures