Learn More
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 396621000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Methyltetrahydrofuran | |
Stabilized | |
Colorless | |
100.0 | |
78.0°C to 80.0°C | |
Authentic | |
Liquid | |
99% min. (GC) | |
C5H10O | |
4 mPa.s (25°C) | |
2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran | |
JWUJQDFVADABEY-UHFFFAOYSA-N | |
2-methyloxolane | |
7301 | |
99+% |
-136.0°C | |
96-47-9 | |
99.0 | |
0.8600g/mL | |
100 mL | |
AcroSeal™ Glass Bottle | |
1.4046 to 1.4066 | |
0.86 | |
MFCD00005367 | |
15,6198 | |
Solubility in water: 150g/l (25°C) | |
CC1CCCO1 | |
86.13 | |
86.13 | |
Extra Dry |
Chemical Identifiers
96-47-9 | |
86.13 | |
JWUJQDFVADABEY-UHFFFAOYSA-N | |
7301 | |
CC1CCCO1 |
C5H10O | |
MFCD00005367 | |
2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran | |
2-methyloxolane |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May form explosive peroxides.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
GHS Signal Word: Danger
EINECSNumber : 202-507-4
RUO – Research Use Only