Learn More
2-(Methylthio)benzoic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1037914
Description
2-(Methylthio)benzoic acid is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(Methylthio)benzoic acid is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.
Solubility
Soluble in Methanol.
Notes
Incompatible with strong oxidizing agents, acids, bases, amines. Store in a cool, dry place. Store in a tightly closed container.
Specifications
2- (Methylthio)benzoic acid | |
166°C to 168°C | |
MFCD00029934 | |
2-methylthio benzoic acid, benzoic acid, 2-methylthio, o-methylthio benzoic acid, benzoic acid, o-methylthio, 2-methylsulfanyl benzoic acid, 2-methylmercaptobenzoic acid, 2-carboxyphenyl methyl sulfide, acide methyl-s-2-benzoique, 2-methylmercapto benzoic acid, acide methyl-s-2-benzoique french | |
LWJQGKJCZOGGPJ-UHFFFAOYSA-N | |
2-methylsulfanylbenzoic acid | |
19498 | |
≥98% |
3724-10-5 | |
C8H8O2S | |
25 g | |
Soluble in Methanol. | |
CSC1=CC=CC=C1C(=O)O | |
168.21 | |
168.21 |
Chemical Identifiers
3724-10-5 | |
MFCD00029934 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only