Learn More
2-(Methylthio)thiophene, 97%
CAS: 5780-36-9 | C5H6S2 | 130.223 g/mol
$376.01 - $1546.41
Chemical Identifiers
| CAS | 5780-36-9 |
|---|---|
| Molecular Formula | C5H6S2 |
| Molecular Weight (g/mol) | 130.223 |
| MDL Number | MFCD00052382 |
| InChI Key | ZLSMPEVZXWDWEK-UHFFFAOYSA-N |
| PubChem CID | 79844 |
| IUPAC Name | 2-methylsulfanylthiophene |
| SMILES | CSC1=CC=CS1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1176306
|
Thermo Scientific Chemicals
A1176306 |
5 g |
Each for $376.01
|
|
|||||
|
AAA1176314
|
Thermo Scientific Chemicals
A1176314 |
25 g |
Each for $1,546.41
|
|
|||||
Description
2-(Methylthio)thiophene is used as an intermediate in synthetic chemistry. It is also used in the preparation of 2-thiocyanato-thiophene by reaction with cyanogen iodide using sodium methylmercatide as a reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(Methylthio)thiophene is used as an intermediate in synthetic chemistry. It is also used in the preparation of 2-thiocyanato-thiophene by reaction with cyanogen iodide using sodium methylmercatide as a reagent.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
| 5780-36-9 | |
| 130.223 | |
| ZLSMPEVZXWDWEK-UHFFFAOYSA-N | |
| 2-methylsulfanylthiophene |
| C5H6S2 | |
| MFCD00052382 | |
| 79844 | |
| CSC1=CC=CS1 |
Specifications
| 5780-36-9 | |
| Stench | |
| 1.596 | |
| 5 g | |
| 106272 | |
| ZLSMPEVZXWDWEK-UHFFFAOYSA-N | |
| 2-methylsulfanylthiophene | |
| 79844 | |
| 97% |
| 82°C to 84°C (15 mmHg) | |
| C5H6S2 | |
| MFCD00052382 | |
| UN3334 | |
| 2-methylthio thiophene, thiophene, 2-methylthio, 2-methylsulfanyl thiophene, 2-methylthiothiophene, 2-mehtylthio thiophene, thiophene,2-methylthio, acmc-20aomi, 2-methylthio-thiophene, 2-methylmercaptothiophene, 2-methylsulfanyl thiophene # | |
| CSC1=CC=CS1 | |
| 130.223 | |
| 130.23 | |
| 2- (Methylthio)thiophene |
Safety and Handling
GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only