Learn More
2-Methylvaleric acid, 98+%, Thermo Scientific Chemicals
$81.30 - $869.53
Chemical Identifiers
CAS | 97-61-0 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00002671 |
InChI Key | OVBFMEVBMNZIBR-UHFFFAOYSA-N |
Synonym | 2-methylvaleric acid, pentanoic acid, 2-methyl, valeric acid, 2-methyl, methylpropylacetic acid, 2-pentanecarboxylic acid, 2-methyl valeric acid, alpha-methylvaleric acid, pentanoic acid, methyl, kyselina 2-methylvalerova, fema no. 2754 |
PubChem CID | 7341 |
IUPAC Name | 2-methylpentanoic acid |
SMILES | CCCC(C)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1387322
|
Thermo Scientific Chemicals
A1387322 |
100 g |
Each for $81.30
|
|
|||||
AAA138730E
|
Thermo Scientific Chemicals
A138730E |
2500 g |
Each for $869.53
|
|
|||||
Description
(R,S)-2-methylvaleric acid is an intermediate in the synthesis of branched-chain. 2-Methylpentanoic acid (2-Methylvaleric acid) was used as an internal standard for gas chromatographic analysis of microbial end products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(R,S)-2-methylvaleric acid is an intermediate in the synthesis of branched-chain. 2-Methylpentanoic acid (2-Methylvaleric acid) was used as an internal standard for gas chromatographic analysis of microbial end products.
Solubility
Soluble in water(13g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Chemical Identifiers
97-61-0 | |
116.16 | |
OVBFMEVBMNZIBR-UHFFFAOYSA-N | |
7341 | |
CCCC(C)C(=O)O |
C6H12O2 | |
MFCD00002671 | |
2-methylvaleric acid, pentanoic acid, 2-methyl, valeric acid, 2-methyl, methylpropylacetic acid, 2-pentanecarboxylic acid, 2-methyl valeric acid, alpha-methylvaleric acid, pentanoic acid, methyl, kyselina 2-methylvalerova, fema no. 2754 | |
2-methylpentanoic acid |
Specifications
97-61-0 | |
0.93 | |
91°C (195°F) | |
C6H12O2 | |
MFCD00002671 | |
UN3265 | |
2-methylvaleric acid, pentanoic acid, 2-methyl, valeric acid, 2-methyl, methylpropylacetic acid, 2-pentanecarboxylic acid, 2-methyl valeric acid, alpha-methylvaleric acid, pentanoic acid, methyl, kyselina 2-methylvalerova, fema no. 2754 | |
OVBFMEVBMNZIBR-UHFFFAOYSA-N | |
2-methylpentanoic acid | |
7341 | |
≥98% |
-85°C | |
196°C to 197°C | |
Unpleasant | |
1.414 | |
100 g | |
1720655 | |
Soluble in water(13g/L). | |
CCCC(C)C(=O)O | |
116.16 | |
116.16 | |
2-Methylvaleric acid |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 202-594-9
RTECSNumber : YV7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only