missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-n-Hexyldecanoic acid, 97%
CAS: 25354-97-6 | C16H32O2 | 256.43 g/mol
$55.78 - $132.40
Chemical Identifiers
| CAS | 25354-97-6 |
|---|---|
| Molecular Formula | C16H32O2 |
| Molecular Weight (g/mol) | 256.43 |
| MDL Number | MFCD00059651 |
| InChI Key | JMOLZNNXZPAGBH-UHFFFAOYSA-N |
| Synonym | decanoic acid, 2-hexyl, hexyldecanoic acid, c16 guerbet fatty acid, 2-hexyldecansaeure, 7-pentadecanecarboxylic acid, 2-hexyldecansaeure german, 2-hexyl-decanoic acid, 2-n-hexyldecanoic acid, 2-hexyldecanoicacid, isocarb 16 |
| PubChem CID | 32912 |
| IUPAC Name | 2-hexyldecanoic acid |
| SMILES | CCCCCCCCC(CCCCCC)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1223814
|
Thermo Scientific Chemicals
L1223814 |
25 g |
Each for $55.78
|
|
|||||
|
AAL1223822
|
Thermo Scientific Chemicals
L1223822 |
100 g |
Each for $132.40
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 25354-97-6 | |
| 256.43 | |
| JMOLZNNXZPAGBH-UHFFFAOYSA-N | |
| 32912 | |
| CCCCCCCCC(CCCCCC)C(=O)O |
| C16H32O2 | |
| MFCD00059651 | |
| decanoic acid, 2-hexyl, hexyldecanoic acid, c16 guerbet fatty acid, 2-hexyldecansaeure, 7-pentadecanecarboxylic acid, 2-hexyldecansaeure german, 2-hexyl-decanoic acid, 2-n-hexyldecanoic acid, 2-hexyldecanoicacid, isocarb 16 | |
| 2-hexyldecanoic acid |
Specifications
| 25354-97-6 | |
| 0.88 | |
| 174°C (345°F) | |
| 1.446 | |
| 25 g | |
| 14,4709 | |
| JMOLZNNXZPAGBH-UHFFFAOYSA-N | |
| 2-hexyldecanoic acid | |
| 32912 | |
| 97% |
| 10°C | |
| 165°C to 168°C (2 mmHg) | |
| C16H32O2 | |
| MFCD00059651 | |
| 1777217 | |
| decanoic acid, 2-hexyl, hexyldecanoic acid, c16 guerbet fatty acid, 2-hexyldecansaeure, 7-pentadecanecarboxylic acid, 2-hexyldecansaeure german, 2-hexyl-decanoic acid, 2-n-hexyldecanoic acid, 2-hexyldecanoicacid, isocarb 16 | |
| CCCCCCCCC(CCCCCC)C(=O)O | |
| 256.43 | |
| 256.43 | |
| 2-n-Hexyldecanoic acid |
Safety and Handling
EINECSNumber : 246-885-9
RTECSNumber : HD9750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only