Learn More
2-(N,N-Dimethylaminomethyl)phenylboronic acid, 98%, Thermo Scientific Chemicals
$299.07 - $299.07
Chemical Identifiers
| CAS | 85107-53-5 |
|---|---|
| Molecular Formula | C9H14BNO2 |
| Molecular Weight (g/mol) | 179.02 |
| MDL Number | MFCD01318999 |
| InChI Key | VUDCTLOJEPCNRS-UHFFFAOYSA-N |
| Synonym | 2-dimethylamino methyl phenyl boronic acid, 2-n,n-dimethylaminomethyl phenylboronic acid, 2-dimethylamino methyl phenylboronic acid, boronic acid, 2-dimethylamino methyl phenyl, 2-n,n-dimethylaminomethyl benzeneboronic acid, 2-n,n-dimethylaminomethyl phenyl-boronic acid, 2-dimethylamino methyl phenyl boranediol, 2-dimethylamino methyl benzeneboronic acid |
| PubChem CID | 2734345 |
| IUPAC Name | [2-[(dimethylamino)methyl]phenyl]boronic acid |
| SMILES | B(C1=CC=CC=C1CN(C)C)(O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC359241000
|
Thermo Scientific Chemicals
359241000 |
100 mg | Glass bottle |
Each for $299.07
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 85107-53-5 | |
| 179.02 | |
| VUDCTLOJEPCNRS-UHFFFAOYSA-N | |
| 2734345 | |
| B(C1=CC=CC=C1CN(C)C)(O)O |
| C9H14BNO2 | |
| MFCD01318999 | |
| 2-dimethylamino methyl phenyl boronic acid, 2-n,n-dimethylaminomethyl phenylboronic acid, 2-dimethylamino methyl phenylboronic acid, boronic acid, 2-dimethylamino methyl phenyl, 2-n,n-dimethylaminomethyl benzeneboronic acid, 2-n,n-dimethylaminomethyl phenyl-boronic acid, 2-dimethylamino methyl phenyl boranediol, 2-dimethylamino methyl benzeneboronic acid | |
| [2-[(dimethylamino)methyl]phenyl]boronic acid |
Specifications
| 85107-53-5 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| MFCD01318999 | |
| 2-dimethylamino methyl phenyl boronic acid, 2-n,n-dimethylaminomethyl phenylboronic acid, 2-dimethylamino methyl phenylboronic acid, boronic acid, 2-dimethylamino methyl phenyl, 2-n,n-dimethylaminomethyl benzeneboronic acid, 2-n,n-dimethylaminomethyl phenyl-boronic acid, 2-dimethylamino methyl phenyl boranediol, 2-dimethylamino methyl benzeneboronic acid | |
| B(C1=CC=CC=C1CN(C)C)(O)O | |
| 179.02 | |
| 179.02 | |
| Powder |
| 97.5 | |
| Beige | |
| 97.5% min. (HPLC) | |
| C9H14BNO2 | |
| 100 mg | |
| VUDCTLOJEPCNRS-UHFFFAOYSA-N | |
| [2-[(dimethylamino)methyl]phenyl]boronic acid | |
| 2734345 | |
| 98% | |
| 2-(N, N-Dimethylaminomethyl)phenylboronic acid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
TSCA : TSCA
RUO – Research Use Only