missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Naphthalenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 158440250

 View more versions of this product

Catalog No. AC158440250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Naphthalenesulfonyl chloride
96.0
74°C to 78°C
Authentic
Glass bottle
C10H7SO2Cl
25 g
2-naphthalenesulfonyl chloride, 2-naphthylsulfonyl chloride, beta-naphthalenesulfonyl chloride, naphthalene-2-sulfonic acid chloride, .beta.-naphthalenesulfochloride, naphthalene-2-sulphonyl chloride, 2-naphthalenesulfonylchloride, 2-naphthalenylsulfonyl chloride, 2-naphthalene sulfonyl chloride, .beta.-naphthalenesulfonyl chloride
OPECTNGATDYLSS-UHFFFAOYSA-N
naphthalene-2-sulfonyl chloride
7125
97%
93-11-8
100.0
Yellow to Beige
97%
C10H7ClO2S
MFCD00004087
11, 173
Solubility in water: insoluble
ClS(=O)(=O)C1=CC=C2C=CC=CC2=C1
226.67
226.68
Powder

Chemical Identifiers

93-11-8
226.67
OPECTNGATDYLSS-UHFFFAOYSA-N
7125
ClS(=O)(=O)C1=CC=C2C=CC=CC2=C1
C10H7ClO2S
MFCD00004087
2-naphthalenesulfonyl chloride, 2-naphthylsulfonyl chloride, beta-naphthalenesulfonyl chloride, naphthalene-2-sulfonic acid chloride, .beta.-naphthalenesulfochloride, naphthalene-2-sulphonyl chloride, 2-naphthalenesulfonylchloride, 2-naphthalenylsulfonyl chloride, 2-naphthalene sulfonyl chloride, .beta.-naphthalenesulfonyl chloride
naphthalene-2-sulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-219-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only