missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Naphthoic acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 177320250

 View more versions of this product

Catalog No. AC177320250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Naphthoic acid
98.5
183°C to 187°C
205g/mL
205°C
98.5% min. (HPLC)
C11H8O2
MFCD00004101
09, 656
2-naphthoic acid, 2-naphthalenecarboxylic acid, isonaphthoic acid, 2-carboxynaphthalene, 2-maythic acid, beta-naphthoic acid, ne-2-carboxylic acid, unii-qlg01v0w2l, naphthalene-beta-carboxylic acid, .beta.-naphthoic acid
UOBYKYZJUGYBDK-UHFFFAOYSA-N
naphthalene-2-carboxylic acid
7123
172.18
Powder
93-09-4
100.0
White to Beige
>300°C
Authentic
Glass bottle
C10H7CO2H
25 g
15, 6467
Solubility in water: <0.5g/L (20°C). Other solubilities: soluble in alcohol and ether
C1=CC=C2C=C(C=CC2=C1)C(=O)O
172.18
CHEBI:36106
99%

Chemical Identifiers

93-09-4
172.18
UOBYKYZJUGYBDK-UHFFFAOYSA-N
7123
naphthalene-2-carboxylic acid
C11H8O2
MFCD00004101
2-naphthoic acid, 2-naphthalenecarboxylic acid, isonaphthoic acid, 2-carboxynaphthalene, 2-maythic acid, beta-naphthoic acid, ne-2-carboxylic acid, unii-qlg01v0w2l, naphthalene-beta-carboxylic acid, .beta.-naphthoic acid
CHEBI:36106
C1=CC=C2C=C(C=CC2=C1)C(=O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 202-217-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only