missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitro-1,4-phenylenediamine 94.0+%, TCI America™

$57.27 - $349.57

Chemical Identifiers

CAS 5307-14-2
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00007903
InChI Key HVHNMNGARPCGGD-UHFFFAOYSA-N
Synonym 2-nitro-1,4-phenylenediamine, 1,4-diamino-2-nitrobenzene, 2-nitro-p-phenylenediamine, 2,5-diaminonitrobenzene, 1,4-benzenediamine, 2-nitro, durafur brown, 2-nitro-1,4-benzenediamine, o-nitro-p-phenylenediamine, 4-amino-2-nitroaniline, fouramine 2r
PubChem CID 4338370
ChEBI CHEBI:76394
IUPAC Name 2-nitrobenzene-1,4-diamine
SMILES C1=CC(=C(C=C1N)[N+](=O)[O-])N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D010525G
View Documents
TCI America
D010525G
25 g
Each for $57.27
 
D0105500G
View Documents
TCI America
D0105500G
500 g
Each for $349.57
 
Description

Description

Specifications

Chemical Identifiers

5307-14-2
153.141
HVHNMNGARPCGGD-UHFFFAOYSA-N
4338370
2-nitrobenzene-1,4-diamine
C6H7N3O2
MFCD00007903
2-nitro-1,4-phenylenediamine, 1,4-diamino-2-nitrobenzene, 2-nitro-p-phenylenediamine, 2,5-diaminonitrobenzene, 1,4-benzenediamine, 2-nitro, durafur brown, 2-nitro-1,4-benzenediamine, o-nitro-p-phenylenediamine, 4-amino-2-nitroaniline, fouramine 2r
CHEBI:76394
C1=CC(=C(C=C1N)[N+](=O)[O-])N

Specifications

5307-14-2
Black-Green
MFCD00007903
2-nitro-1,4-phenylenediamine, 1,4-diamino-2-nitrobenzene, 2-nitro-p-phenylenediamine, 2,5-diaminonitrobenzene, 1,4-benzenediamine, 2-nitro, durafur brown, 2-nitro-1,4-benzenediamine, o-nitro-p-phenylenediamine, 4-amino-2-nitroaniline, fouramine 2r
C1=CC(=C(C=C1N)[N+](=O)[O-])N
153.141
CHEBI:76394
≥94.0% (T)
2-Nitro-1,4-phenylenediamine
138°C
C6H7N3O2
25 g
HVHNMNGARPCGGD-UHFFFAOYSA-N
2-nitrobenzene-1,4-diamine
4338370
153.14
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0402

RTECSNumber : ST3000000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'