missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-(trifluoromethyl)aniline, 98%

CAS: 400-98-6 | C7H5F3N2O2 | 206.124 g/mol

$78.03 - $812.04

Chemical Identifiers

CAS 400-98-6
Molecular Formula C7H5F3N2O2
Molecular Weight (g/mol) 206.124
MDL Number MFCD00007155
InChI Key ATXBGHLILIABGX-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethyl aniline, 4-amino-3-nitrobenzotrifluoride, benzenamine, 2-nitro-4-trifluoromethyl, alpha,alpha,alpha-trifluoro-2-nitro-p-toluidine, 4-trifluoromethyl 2-nitroaniline, 4-amino-3-nitro benzotrifluoride, 4-amino-3-nitrobenzo trifluoride, aniline, 2-nitro-4-trifluoromethyl, 2-nitro-4-trifluoromethyl phenylamine, 3-nitro-4-aminobenzotrifluoride
PubChem CID 67865
IUPAC Name 2-nitro-4-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1441309
View Documents
Thermo Scientific Chemicals
A1441309
10 g
Each for $78.03
Only null left
 
AAA1441318
View Documents
Thermo Scientific Chemicals
A1441318
50 g
Each for $201.25
Only null left
 
AAA1441330
View Documents
Thermo Scientific Chemicals
A1441330
250 g
Each for $812.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

400-98-6
206.124
ATXBGHLILIABGX-UHFFFAOYSA-N
67865
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])N
C7H5F3N2O2
MFCD00007155
2-nitro-4-trifluoromethyl aniline, 4-amino-3-nitrobenzotrifluoride, benzenamine, 2-nitro-4-trifluoromethyl, alpha,alpha,alpha-trifluoro-2-nitro-p-toluidine, 4-trifluoromethyl 2-nitroaniline, 4-amino-3-nitro benzotrifluoride, 4-amino-3-nitrobenzo trifluoride, aniline, 2-nitro-4-trifluoromethyl, 2-nitro-4-trifluoromethyl phenylamine, 3-nitro-4-aminobenzotrifluoride
2-nitro-4-(trifluoromethyl)aniline

Specifications

400-98-6
C7H5F3N2O2
10 g
650808
ATXBGHLILIABGX-UHFFFAOYSA-N
2-nitro-4-(trifluoromethyl)aniline
67865
98%
105°C to 109°C
MFCD00007155
UN2811
2-nitro-4-trifluoromethyl aniline, 4-amino-3-nitrobenzotrifluoride, benzenamine, 2-nitro-4-trifluoromethyl, alpha,alpha,alpha-trifluoro-2-nitro-p-toluidine, 4-trifluoromethyl 2-nitroaniline, 4-amino-3-nitro benzotrifluoride, 4-amino-3-nitrobenzo trifluoride, aniline, 2-nitro-4-trifluoromethyl, 2-nitro-4-trifluoromethyl phenylamine, 3-nitro-4-aminobenzotrifluoride
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])N
206.124
206.13
2-Nitro-4-(trifluoromethyl)aniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 206-926-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only