missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Nitroacetanilide, 98+%, Thermo Scientific Chemicals

$67.58 - $250.19

Chemical Identifiers

CAS 552-32-9
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.16
MDL Number MFCD00016991
InChI Key BUNFNRVLMKHKIT-UHFFFAOYSA-N
Synonym n-2-nitrophenyl acetamide, 2'-nitroacetanilide, o-nitroacetanilide, 2-nitroacetanilide, acetamide, n-2-nitrophenyl, acetanilide, 2'-nitro, unii-q4kjc83992, o-nitroacetoanilide, zlchem 447, pubchem3357
PubChem CID 11090
IUPAC Name N-(2-nitrophenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1667706
View Documents
Thermo Scientific Chemicals
A1667706
5 g
Each for $67.58
 
AAA1667714
View Documents
Thermo Scientific Chemicals
A1667714
25 g
Each for $250.19
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

552-32-9
180.16
BUNFNRVLMKHKIT-UHFFFAOYSA-N
11090
CC(=O)NC1=CC=CC=C1[N+]([O-])=O
C8H8N2O3
MFCD00016991
n-2-nitrophenyl acetamide, 2'-nitroacetanilide, o-nitroacetanilide, 2-nitroacetanilide, acetamide, n-2-nitrophenyl, acetanilide, 2'-nitro, unii-q4kjc83992, o-nitroacetoanilide, zlchem 447, pubchem3357
N-(2-nitrophenyl)acetamide

Specifications

552-32-9
1.42
MFCD00016991
1959178
n-2-nitrophenyl acetamide, 2'-nitroacetanilide, o-nitroacetanilide, 2-nitroacetanilide, acetamide, n-2-nitrophenyl, acetanilide, 2'-nitro, unii-q4kjc83992, o-nitroacetoanilide, zlchem 447, pubchem3357
CC(=O)NC1=CC=CC=C1[N+]([O-])=O
180.16
180.16
2'-Nitroacetanilide
90°C to 93°C
C8H8N2O3
5 g
14,6581
BUNFNRVLMKHKIT-UHFFFAOYSA-N
N-(2-nitrophenyl)acetamide
11090
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-009-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only