missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroaniline, 98%

CAS: 88-74-4 | C6H6N2O2 | 138.13 g/mol

$73.24 - $254.11

Chemical Identifiers

CAS 88-74-4
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00007687
InChI Key DPJCXCZTLWNFOH-UHFFFAOYSA-N
Synonym o-nitroaniline, benzenamine, 2-nitro, o-nitraniline, 1-amino-2-nitrobenzene, 2-nitrobenzenamine, aniline, o-nitro, devol orange b, orange grs salt, o-aminonitrobenzene, 2-aminonitrobenzene
PubChem CID 6946
IUPAC Name 2-nitroaniline
SMILES C1=CC=C(C(=C1)N)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC128351000
SDS View Documents
Thermo Scientific Chemicals
128351000
100 g Glass bottle N/A
Only null left
 
AC128355000
SDS View Documents
Thermo Scientific Chemicals
128355000
500 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

88-74-4
138.13
DPJCXCZTLWNFOH-UHFFFAOYSA-N
6946
C1=CC=C(C(=C1)N)[N+](=O)[O-]
C6H6N2O2
MFCD00007687
o-nitroaniline, benzenamine, 2-nitro, o-nitraniline, 1-amino-2-nitrobenzene, 2-nitrobenzenamine, aniline, o-nitro, devol orange b, orange grs salt, o-aminonitrobenzene, 2-aminonitrobenzene
2-nitroaniline

Specifications

88-74-4
100.0
Orange to Brown
168°C
97.5% min. (GC)
C6H6N2O2
MFCD00007687
12,687
o-nitroaniline, benzenamine, 2-nitro, o-nitraniline, 1-amino-2-nitrobenzene, 2-nitrobenzenamine, aniline, o-nitro, devol orange b, orange grs salt, o-aminonitrobenzene, 2-aminonitrobenzene
DPJCXCZTLWNFOH-UHFFFAOYSA-N
2-nitroaniline
6946
98%
2-Nitroaniline, 98%
97.5
70.0°C to 74.0°C
284.0°C
Authentic
Glass bottle
O2NC6H4NH2
100 g
15,6669
Solubility in water: 1.1g/L in water (20°C). Other solubilities: soluble in alcohol,chloroform and ether
C1=CC=C(C(=C1)N)[N+](=O)[O-]
138.13
138.13
Crystals or Flakes
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Harmful to aquatic life with long lasting effects.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-855-4

RTECSNumber : BY6650000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only