Learn More
2-Nitrobenzoic acid, 95%, Thermo Scientific Chemicals
$72.33 - $734.52
Chemical Identifiers
CAS | 552-16-9 |
---|---|
Molecular Formula | C7H5NO4 |
Molecular Weight (g/mol) | 167.12 |
MDL Number | MFCD00007137 |
InChI Key | SLAMLWHELXOEJZ-UHFFFAOYSA-N |
Synonym | 2-nitrobenzoic acid, o-nitrobenzoic acid, benzoic acid, 2-nitro, 2-nitrobenzoicacid, o-carboxynitrobenzene, benzoic acid, o-nitro, nitrobenzoic acid, 2-nitro-benzoic acid, ccris 2334, 2-nitro benzoic acid |
PubChem CID | 11087 |
ChEBI | CHEBI:25620 |
SMILES | C1=CC=C(C(=C1)C(=O)O)[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1039322
|
Thermo Scientific Chemicals
A1039322 |
100 g |
Each for $72.33
|
|
|||||
AAA1039336
|
Thermo Scientific Chemicals
A1039336 |
500 g |
Each for $230.10
|
|
|||||
AAA103930E
|
Thermo Scientific Chemicals
A103930E |
2500 g |
Each for $734.52
|
|
|||||
Description
It is employed as a synthetic reagent and can function as a growth supplement for Pseudomonas fluorescens strain KU-7. It also finds its use as a reagent for protection of amine groups and in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a synthetic reagent and can function as a growth supplement for Pseudomonas fluorescens strain KU-7. It also finds its use as a reagent for protection of amine groups and in organic synthesis.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
552-16-9 | |
167.12 | |
SLAMLWHELXOEJZ-UHFFFAOYSA-N | |
11087 | |
C1=CC=C(C(=C1)C(=O)O)[N+](=O)[O-] |
C7H5NO4 | |
MFCD00007137 | |
2-nitrobenzoic acid, o-nitrobenzoic acid, benzoic acid, 2-nitro, 2-nitrobenzoicacid, o-carboxynitrobenzene, benzoic acid, o-nitro, nitrobenzoic acid, 2-nitro-benzoic acid, ccris 2334, 2-nitro benzoic acid | |
CHEBI:25620 |
Specifications
552-16-9 | |
1.58 | |
MFCD00007137 | |
1910632 | |
2-nitrobenzoic acid, o-nitrobenzoic acid, benzoic acid, 2-nitro, 2-nitrobenzoicacid, o-carboxynitrobenzene, benzoic acid, o-nitro, nitrobenzoic acid, 2-nitro-benzoic acid, ccris 2334, 2-nitro benzoic acid | |
SLAMLWHELXOEJZ-UHFFFAOYSA-N | |
167.12 | |
CHEBI:25620 | |
95% |
146°C to 149°C | |
C7H5NO4 | |
100 g | |
14,6589 | |
Soluble in water. | |
C1=CC=C(C(=C1)C(=O)O)[N+](=O)[O-] | |
11087 | |
167.12 | |
2-Nitrobenzoic acid |
Safety and Handling
GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-004-9
RTECSNumber : DH5050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only