Learn More
2-Nitrobenzoyl chloride, 95%, Thermo Scientific Chemicals
$220.34 - $220.34
Chemical Identifiers
CAS | 610-14-0 |
---|---|
Molecular Formula | C7H4ClNO3 |
Molecular Weight (g/mol) | 185.57 |
MDL Number | MFCD00007131 |
InChI Key | BWWHTIHDQBHTHP-UHFFFAOYSA-N |
Synonym | o-nitrobenzoyl chloride, benzoyl chloride, 2-nitro, 2-nitrobenzoylchloride, benzoyl chloride, o-nitro, unii-86w2g0ja5q, nitrobenzoyl chloride, ccris 3135, 2-nitro-benzoyl chloride, benzoyl chloride, nitro, dsstox_cid_5741 |
PubChem CID | 11875 |
IUPAC Name | 2-nitrobenzoyl chloride |
SMILES | C1=CC=C(C(=C1)C(=O)Cl)[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC232310250
|
Thermo Scientific Chemicals
232310250 |
25 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
610-14-0 | |
185.57 | |
BWWHTIHDQBHTHP-UHFFFAOYSA-N | |
11875 | |
C1=CC=C(C(=C1)C(=O)Cl)[N+](=O)[O-] |
C7H4ClNO3 | |
MFCD00007131 | |
o-nitrobenzoyl chloride, benzoyl chloride, 2-nitro, 2-nitrobenzoylchloride, benzoyl chloride, o-nitro, unii-86w2g0ja5q, nitrobenzoyl chloride, ccris 3135, 2-nitro-benzoyl chloride, benzoyl chloride, nitro, dsstox_cid_5741 | |
2-nitrobenzoyl chloride |
Specifications
610-14-0 | |
100.0 | |
Brown to Yellow | |
148°C to 149°C (9.0 mmHg) | |
95% | |
C7H4ClNO3 | |
O2NC6H4COCl | |
25 g | |
o-nitrobenzoyl chloride, benzoyl chloride, 2-nitro, 2-nitrobenzoylchloride, benzoyl chloride, o-nitro, unii-86w2g0ja5q, nitrobenzoyl chloride, ccris 3135, 2-nitro-benzoyl chloride, benzoyl chloride, nitro, dsstox_cid_5741 | |
BWWHTIHDQBHTHP-UHFFFAOYSA-N | |
2-nitrobenzoyl chloride | |
11875 | |
95% | |
2-Nitrobenzoyl chloride |
94.0 | |
17°C to 20°C | |
1.4000g/mL | |
Authentic | |
Glass bottle | |
1.5666 to 1.5686 | |
MFCD00007131 | |
1.4 | |
Solubility in water: decomposes | |
C1=CC=C(C(=C1)C(=O)Cl)[N+](=O)[O-] | |
185.57 | |
185.57 | |
Liquid After Melting |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 210-206-4
RUO – Research Use Only