Learn More
2-Nitrobenzyl alcohol, 97%
CAS: 612-25-9 | C7H7NO3 | 153.14 g/mol
$103.19 - $294.47
Chemical Identifiers
| CAS | 612-25-9 |
|---|---|
| Molecular Formula | C7H7NO3 |
| Molecular Weight (g/mol) | 153.14 |
| MDL Number | MFCD00007186 |
| InChI Key | BWRBVBFLFQKBPT-UHFFFAOYSA-N |
| Synonym | 2-nitrobenzyl alcohol, 2-nitrophenyl methanol, o-nitrobenzyl alcohol, benzyl alcohol, o-nitro, benzenemethanol, 2-nitro, 2-nitrobenzenemethanol, o-hydroxymethylnitrobenzene, ccris 7967, benzenemethanol, nitro, 2-nitrophenyl methan-1-ol |
| PubChem CID | 11923 |
| IUPAC Name | (2-nitrophenyl)methanol |
| SMILES | OCC1=CC=CC=C1[N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC128510250
|
Thermo Scientific Chemicals
128510250 |
25 g | Glass Bottle |
Each for $103.19
|
|
||||
|
AC128511000
|
Thermo Scientific Chemicals
128511000 |
100 g | Glass Bottle |
Each for $294.47
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 612-25-9 | |
| 153.14 | |
| BWRBVBFLFQKBPT-UHFFFAOYSA-N | |
| 11923 | |
| OCC1=CC=CC=C1[N+]([O-])=O |
| C7H7NO3 | |
| MFCD00007186 | |
| 2-nitrobenzyl alcohol, 2-nitrophenyl methanol, o-nitrobenzyl alcohol, benzyl alcohol, o-nitro, benzenemethanol, 2-nitro, 2-nitrobenzenemethanol, o-hydroxymethylnitrobenzene, ccris 7967, benzenemethanol, nitro, 2-nitrophenyl methan-1-ol | |
| (2-nitrophenyl)methanol |
Specifications
| 612-25-9 | |
| 100.0 | |
| Beige to Beige-Brown | |
| 97% | |
| C7H7NO3 | |
| MFCD00007186 | |
| 06, 447 | |
| BWRBVBFLFQKBPT-UHFFFAOYSA-N | |
| (2-nitrophenyl)methanol | |
| 11923 | |
| 97% | |
| 2-Nitrobenzyl alcohol, 97% |
| 96.0 | |
| 69°C to 73°C | |
| Authentic | |
| Glass Bottle | |
| O2NC6H4CH2OH | |
| 25 g | |
| 2-nitrobenzyl alcohol, 2-nitrophenyl methanol, o-nitrobenzyl alcohol, benzyl alcohol, o-nitro, benzenemethanol, 2-nitro, 2-nitrobenzenemethanol, o-hydroxymethylnitrobenzene, ccris 7967, benzenemethanol, nitro, 2-nitrophenyl methan-1-ol | |
| OCC1=CC=CC=C1[N+]([O-])=O | |
| 153.14 | |
| 153.14 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 210-302-6
RTECSNumber : DP0657050
TSCA : TSCA
RUO – Research Use Only