Learn More
2-Nitrobenzylamine hydrochloride, 98%, Thermo Scientific Chemicals
$114.32 - $412.71
Chemical Identifiers
CAS | 24835-08-3 |
---|---|
Molecular Formula | C7H9ClN2O2 |
Molecular Weight (g/mol) | 188.611 |
MDL Number | MFCD00136280 |
InChI Key | BASJTVIZZDEQBJ-UHFFFAOYSA-N |
Synonym | 2-nitrobenzylamine hydrochloride, 2-nitrophenyl methanamine hydrochloride, 1-2-nitrophenyl methanamine hydrochloride, 2-nitrobenzylamine hcl, 2-nitrobenzyl amine hcl, acmc-1co7n, o-nitrobenzylamine hydrochloride, 2-nitrobenzyl amine hydrochloride, 2-nitrobenzenemethanamine hydrochloride, 2-nitrophenyl methanaminehydrochloride |
PubChem CID | 12235442 |
IUPAC Name | (2-nitrophenyl)methanamine;hydrochloride |
SMILES | C1=CC=C(C(=C1)CN)[N+](=O)[O-].Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1890003
|
Thermo Scientific Chemicals
A1890003 |
1 g |
Each for $114.32
|
|
|||||
AAA1890006
|
Thermo Scientific Chemicals
A1890006 |
5 g |
Each for $412.71
|
|
|||||
Description
2-Nitrobenzylamine hydrochloride may be used in chemical synthesis studies. Also reactant involved in synthesis of tetrahydroisoquinolines via Heck-aza-Michael reactions, aminomethylphenylamine scaffolds for factor Xa inhibition, squaryliamides for mannosyltransferase inhibition and fluorescent two-photon absorption benzothiadiazole dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Nitrobenzylamine hydrochloride may be used in chemical synthesis studies. Also reactant involved in synthesis of tetrahydroisoquinolines via Heck-aza-Michael reactions, aminomethylphenylamine scaffolds for factor Xa inhibition, squaryliamides for mannosyltransferase inhibition and fluorescent two-photon absorption benzothiadiazole dyes.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
24835-08-3 | |
188.611 | |
BASJTVIZZDEQBJ-UHFFFAOYSA-N | |
12235442 | |
C1=CC=C(C(=C1)CN)[N+](=O)[O-].Cl |
C7H9ClN2O2 | |
MFCD00136280 | |
2-nitrobenzylamine hydrochloride, 2-nitrophenyl methanamine hydrochloride, 1-2-nitrophenyl methanamine hydrochloride, 2-nitrobenzylamine hcl, 2-nitrobenzyl amine hcl, acmc-1co7n, o-nitrobenzylamine hydrochloride, 2-nitrobenzyl amine hydrochloride, 2-nitrobenzenemethanamine hydrochloride, 2-nitrophenyl methanaminehydrochloride | |
(2-nitrophenyl)methanamine;hydrochloride |
Specifications
24835-08-3 | |
C7H9ClN2O2 | |
1 g | |
2-nitrobenzylamine hydrochloride, 2-nitrophenyl methanamine hydrochloride, 1-2-nitrophenyl methanamine hydrochloride, 2-nitrobenzylamine hcl, 2-nitrobenzyl amine hcl, acmc-1co7n, o-nitrobenzylamine hydrochloride, 2-nitrobenzyl amine hydrochloride, 2-nitrobenzenemethanamine hydrochloride, 2-nitrophenyl methanaminehydrochloride | |
C1=CC=C(C(=C1)CN)[N+](=O)[O-].Cl | |
188.611 | |
188.62 | |
2-Nitrobenzylamine hydrochloride |
250°C (decomposition) | |
MFCD00136280 | |
Hygroscopic | |
BASJTVIZZDEQBJ-UHFFFAOYSA-N | |
(2-nitrophenyl)methanamine;hydrochloride | |
12235442 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only