missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitrochalcone, 97%, Thermo Scientific Chemicals

$113.14 - $424.92

Chemical Identifiers

CAS 7473-93-0
Molecular Formula C15H11NO3
Molecular Weight (g/mol) 253.257
MDL Number MFCD00031069
InChI Key KTXHLWZQKQDFRF-ZHACJKMWSA-N
Synonym 2-nitrochalcone, chalcone, 2-nitro, 2-nitrobenzylideneacetophenone, 2e-3-2-nitrophenyl-1-phenylprop-2-en-1-one, ccris 1671, e-3-2-nitro-phenyl-1-phenyl-propenone, 2-propen-1-one, 3-2-nitrophenyl-1-phenyl, e-3-2-nitrophenyl-1-phenyl-2-propen-1-one, e-3-2-nitrophenyl-1-phenylprop-2-en-1-one, e-3-2-nitrophenyl-1-phenyl-prop-2-en-1-one
PubChem CID 5337611
IUPAC Name (E)-3-(2-nitrophenyl)-1-phenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C(=O)C=CC2=CC=CC=C2[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1776306
View Documents
Thermo Scientific Chemicals
A1776306
5 g
Each for $113.14
 
AAA1776314
View Documents
Thermo Scientific Chemicals
A1776314
25 g
Each for $424.92
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7473-93-0
253.257
KTXHLWZQKQDFRF-ZHACJKMWSA-N
5337611
C1=CC=C(C=C1)C(=O)C=CC2=CC=CC=C2[N+](=O)[O-]
C15H11NO3
MFCD00031069
2-nitrochalcone, chalcone, 2-nitro, 2-nitrobenzylideneacetophenone, 2e-3-2-nitrophenyl-1-phenylprop-2-en-1-one, ccris 1671, e-3-2-nitro-phenyl-1-phenyl-propenone, 2-propen-1-one, 3-2-nitrophenyl-1-phenyl, e-3-2-nitrophenyl-1-phenyl-2-propen-1-one, e-3-2-nitrophenyl-1-phenylprop-2-en-1-one, e-3-2-nitrophenyl-1-phenyl-prop-2-en-1-one
(E)-3-(2-nitrophenyl)-1-phenylprop-2-en-1-one

Specifications

7473-93-0
C15H11NO3
5 g
KTXHLWZQKQDFRF-ZHACJKMWSA-N
(E)-3-(2-nitrophenyl)-1-phenylprop-2-en-1-one
5337611
97%
124°C to 128°C
MFCD00031069
2-nitrochalcone, chalcone, 2-nitro, 2-nitrobenzylideneacetophenone, 2e-3-2-nitrophenyl-1-phenylprop-2-en-1-one, ccris 1671, e-3-2-nitro-phenyl-1-phenyl-propenone, 2-propen-1-one, 3-2-nitrophenyl-1-phenyl, e-3-2-nitrophenyl-1-phenyl-2-propen-1-one, e-3-2-nitrophenyl-1-phenylprop-2-en-1-one, e-3-2-nitrophenyl-1-phenyl-prop-2-en-1-one
C1=CC=C(C=C1)C(=O)C=CC2=CC=CC=C2[N+](=O)[O-]
253.257
253.26
2-Nitrochalcone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only