missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitrotoluene, 99+%, Thermo Scientific Chemicals

Used as an intermediate, reagent, as a precursor and as a nitrogen, carbon, energy supplement

Supplier:  Thermo Scientific Chemicals A1344636

 View more versions of this product

Catalog No. AAA1344636


Explore available promotions

Description

Description

2-Nitrotoluene is used as an intermediate for the o-toludine synthesis, which finds application in the dyestuff industry. It is employed as a reagent in detecting and photometric determining oxidizing agent. It is also used in the determination of prussic acid content in the air. It acts as a precursor to prepare 2-amino-4-chlorotoluene, 2-amino-6-chlorotoluene, nitrotoluenesulfonic acids and toluene nitrosulfonic chlorides. It acts as a nitrogen supplement in the culture medium of pseudomonas sp. strain ClS1. It acts as a carbon and energy supplement in the culture medium of Pseudomonas sp. strain JS42.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrotoluene is used as an intermediate for the o-toludine synthesis, which finds application in the dyestuff industry. It is employed as a reagent in detecting and photometric determining oxidizing agent. It is also used in the determination of prussic acid content in the air. It acts as a precursor to prepare 2-amino-4-chlorotoluene, 2-amino-6-chlorotoluene, nitrotoluenesulfonic acids and toluene nitrosulfonic chlorides. It acts as a nitrogen supplement in the culture medium of pseudomonas sp. strain ClS1. It acts as a carbon and energy supplement in the culture medium of Pseudomonas sp. strain JS42.

Solubility
Miscible with acetone, benzene, chloroform, diethyl ether, ethanol and petroleum ether. Immiscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases, sulfuric acid, reducing agents, hydrogen and sodium.
Specifications

Specifications

2-Nitrotoluene
-4°C
224°C to 226°C
C7H7NO2
MFCD00007157
UN1664
14,6650
Miscible with acetone,benzene,chloroform,diethyl ether,ethanol and petroleum ether; Immiscible with water.
CC1=CC=CC=C1[N+](=O)[O-]
137.138
CHEBI:33098
≥99%
88-72-2
1.166
106°C (222°F)
1.545
500 g
1907580
2-nitrotoluene, o-nitrotoluene, o-nitrotoluol, o-methylnitrobenzene, benzene, 1-methyl-2-nitro, toluene, o-nitro, 2-methyl-1-nitrobenzene, 2-nitrotoluol, ortho-nitrotoluol, 2-methylnitrobenzene
PLAZTCDQAHEYBI-UHFFFAOYSA-N
1-methyl-2-nitrobenzene
6944
137.14

Chemical Identifiers

88-72-2
137.138
PLAZTCDQAHEYBI-UHFFFAOYSA-N
6944
1-methyl-2-nitrobenzene
C7H7NO2
MFCD00007157
2-nitrotoluene, o-nitrotoluene, o-nitrotoluol, o-methylnitrobenzene, benzene, 1-methyl-2-nitro, toluene, o-nitro, 2-methyl-1-nitrobenzene, 2-nitrotoluol, ortho-nitrotoluol, 2-methylnitrobenzene
CHEBI:33098
CC1=CC=CC=C1[N+](=O)[O-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H340-H350-H361-H302
May cause genetic defects.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.

P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H340-H350-H361f

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROTOLUENES, LIQUID

EINECSNumber : 201-853-3

RTECSNumber : XT3150000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only