missing translation for 'onlineSavingsMsg'
Learn More

2-Nonadecanone, tech. 80%

CAS: 629-66-3 | C19H38O | 282.51 g/mol

$115.79 - $423.67

Chemical Identifiers

CAS 629-66-3
Molecular Formula C19H38O
Molecular Weight (g/mol) 282.51
MDL Number MFCD00026671
InChI Key IEDKVDCIEARIIU-UHFFFAOYSA-N
Synonym 2-nonadecanone, nonadeca-2-one, methyl heptadecyl ketone, acmc-20anop, 2-oxononadecan-3-yl, n-heptadecyl methyl ketone
PubChem CID 69423
IUPAC Name nonadecan-2-one
SMILES CCCCCCCCCCCCCCCCCC(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2117806
View Documents
Thermo Scientific Chemicals
B2117806
5 g N/A
Only null left
 
AAB2117814
View Documents
Thermo Scientific Chemicals
B2117814
25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

629-66-3
282.51
IEDKVDCIEARIIU-UHFFFAOYSA-N
69423
CCCCCCCCCCCCCCCCCC(C)=O
C19H38O
MFCD00026671
2-nonadecanone, nonadeca-2-one, methyl heptadecyl ketone, acmc-20anop, 2-oxononadecan-3-yl, n-heptadecyl methyl ketone
nonadecan-2-one

Specifications

629-66-3
>100°C (212°F)
CH3(CH2)16COCH3
5 g
2-nonadecanone, nonadeca-2-one, methyl heptadecyl ketone, acmc-20anop, 2-oxononadecan-3-yl, n-heptadecyl methyl ketone
CCCCCCCCCCCCCCCCCC(C)=O
282.51
282.51
Technical
54°C to 56°C
C19H38O
MFCD00026671
1708703
IEDKVDCIEARIIU-UHFFFAOYSA-N
nonadecan-2-one
69423
80%
2-Nonadecanone
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-102-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only