missing translation for 'onlineSavingsMsg'
Learn More

2-Nonyn-1-ol, 96%

CAS: 5921-73-3 | C9H16O | 140.23 g/mol

$168.42 - $621.38

Chemical Identifiers

CAS 5921-73-3
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00041566
InChI Key XERDOEKKHDALKJ-UHFFFAOYSA-N
Synonym 2-nonyn-1-ol, pubchem13067, acmc-1apv0
PubChem CID 80017
IUPAC Name non-2-yn-1-ol
SMILES CCCCCCC#CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0638006
View Documents
Thermo Scientific Chemicals
L0638006
5 g N/A
Only null left
 
AAL0638014
View Documents
Thermo Scientific Chemicals
L0638014
25 g N/A
Only null left
 
Description

Description

2-Nonyn-1-ol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nonyn-1-ol is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

5921-73-3
140.23
XERDOEKKHDALKJ-UHFFFAOYSA-N
80017
CCCCCCC#CCO
C9H16O
MFCD00041566
2-nonyn-1-ol, pubchem13067, acmc-1apv0
non-2-yn-1-ol

Specifications

5921-73-3
109°C to 111°C (11 mmHg)
1.456
5 g
2-nonyn-1-ol, pubchem13067, acmc-1apv0
XERDOEKKHDALKJ-UHFFFAOYSA-N
non-2-yn-1-ol
80017
96%
0.873
C9H16O
MFCD00041566
1745753
Not miscible in water.
CCCCCCC#CCO
140.23
140.23
2-Nonyn-1-ol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-646-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only