missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Octanone, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 129420250

 View more versions of this product

Catalog No. AC129420250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Octanone
111-13-7
100.0
Colorless to Yellow
173°C
Authentic
Glass bottle
1.4140 to 1.416
MFCD00009540
01, 704
15, 4747
Solubility in water: 0.9g/L. Other solubilities: miscible with alcohol,ether,hydrocarbons and,esters
CCCCCCC(=O)C
128.21
CHEBI:87434
99+%
99+%
99.0
-16°C
0.8190g/mL
55°C
99% min. (GC)
C8H16O
CH3(CH2)5COCH3
25 mL
0.819
2-octanone, n-hexyl methyl ketone, hexyl methyl ketone, methyl hexyl ketone, methyl n-hexyl ketone, octanone, 2-oxooctane, 2-octanone natural, fema number 2802, unii-j2g84h29af
ZPVFWPFBNIEHGJ-UHFFFAOYSA-N
octan-2-one
8093
128.21
Liquid

Chemical Identifiers

111-13-7
ZPVFWPFBNIEHGJ-UHFFFAOYSA-N
8093
octan-2-one
MFCD00009540
2-octanone, n-hexyl methyl ketone, hexyl methyl ketone, methyl hexyl ketone, methyl n-hexyl ketone, octanone, 2-oxooctane, 2-octanone natural, fema number 2802, unii-j2g84h29af
CHEBI:87434
CCCCCCC(=O)C
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-837-1

RTECSNumber : RH1484000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only