missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Octanone oxime, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 390280250

 View more versions of this product

Catalog No. AC390280250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2-Octanone oxime
98.5
0.8800g/mL
98.5% min. (GC)
C8H17NO
CH3(CH2)5C(CH3)=NOH
25g
2-octanone, oxime, 2-octanone oxime, n-octan-2-ylidene hydroxylamine, 2-hydroxyiminooctane, z-n-octan-2-ylidene hydroxylamine
CCCCCCC(=NO)C
143.23
143.23
7207-49-0
100.0
Authentic
Glass bottle
1.4510 to 1.453
MFCD00089167
0.88
GZRPVYSKBVDCBV-CMDGGOBGSA-N
(NE)-N-octan-2-ylidenehydroxylamine
9562584
99%

Chemical Identifiers

7207-49-0
143.23
GZRPVYSKBVDCBV-CMDGGOBGSA-N
9562584
CCCCCCC(=NO)C
C8H17NO
MFCD00089167
2-octanone, oxime, 2-octanone oxime, n-octan-2-ylidene hydroxylamine, 2-hydroxyiminooctane, z-n-octan-2-ylidene hydroxylamine
(NE)-N-octan-2-ylidenehydroxylamine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty o

GHS Signal Word: Warning

EINECSNumber : 230-586-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only