missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Oxa-6-azaspiro[3.4]octane, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H57579ME

 View more versions of this product

Catalog No. AAH57579ME


Explore available promotions

Description

Description

2-Oxa-6-azaspiro[3.4]octane is involved in the preparation of azaspirocycle or azetidine substituted 4-anilinoquinazoline derivatives, which exhibits epidermal growth factor receptor (EGFR) inhibitory activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Oxa-6-azaspiro[3.4]octane is involved in the preparation of azaspirocycle or azetidine substituted 4-anilinoquinazoline derivatives, which exhibits epidermal growth factor receptor (EGFR) inhibitory activities.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids, reducing agents and bases.
Specifications

Specifications

2-Oxa-6-azaspiro[3.4]octane
1.08
63°C (145°F)
MFCD14586455
2-oxa-6-azaspiro 3.4 octane, 2-oxa-6-aza-spiro 3.4 octane, 2-oxa-6-azaspiro 3,4 octane, acmc-209fqv, 2-oxa-7-azaspiro 3.4 octane, 2-oxa-6-aza-spiro 3,4 octane, spiro pyrrolidine-3,3'-oxetane
ZHAIMJRKJKQNQI-UHFFFAOYSA-N
2-oxa-7-azaspiro[3.4]octane
53438323
95%
220290-68-6
187°C
C6H11NO
500 mg
Soluble in water.
C1CNCC12COC2
113.16
113.16

Chemical Identifiers

220290-68-6
113.16
ZHAIMJRKJKQNQI-UHFFFAOYSA-N
53438323
C1CNCC12COC2
C6H11NO
MFCD14586455
2-oxa-6-azaspiro 3.4 octane, 2-oxa-6-aza-spiro 3.4 octane, 2-oxa-6-azaspiro 3,4 octane, acmc-209fqv, 2-oxa-7-azaspiro 3.4 octane, 2-oxa-6-aza-spiro 3,4 octane, spiro pyrrolidine-3,3'-oxetane
2-oxa-7-azaspiro[3.4]octane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only