missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Oxa-7-azaspiro[3.5]nonane hemioxalate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H57834ME

 View more versions of this product

Catalog No. AAH57834ME


Explore available promotions

Description

Description

Spiro forms of oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photo chromic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Spiro forms of oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photo chromic materials.

Notes
Hygroscopic. Store at room temperature.
Specifications

Specifications

2-Oxa-7-azaspiro[3.5]nonane hemioxalate
208°C
1.048 g/mL
MFCD11035888
2-oxa-7-azaspiro 3.5 nonane oxalate 2:1, 2-oxa-7-azaspiro 3.5 nonane hemioxalate, bis 2-oxa-7-azaspiro 3.5 nonane ; oxalic acid, 2-oxa-7-azaspiro 3.5 nonan hemioxalate, 2-oxa-7-azaspiro 3,5 nonane hemioxalate, bis 2-oxa-7-azaspiro 3.5 nonane oxalate, 2-oxa-7-azaspiro 3.5 nonane hemi-oxalate
C1CNCCC12COC2.C1CNCCC12COC2.C(=O)(C(=O)O)O
344.408
172.2
Solid
1429056-28-9
White to Gray
C16H28N2O6
500 mg
WWVUFRRXXSVWBJ-UHFFFAOYSA-N
2-oxa-7-azaspiro[3.5]nonane;oxalic acid
71463619
97%

Chemical Identifiers

1429056-28-9
344.408
WWVUFRRXXSVWBJ-UHFFFAOYSA-N
71463619
C1CNCCC12COC2.C1CNCCC12COC2.C(=O)(C(=O)O)O
C16H28N2O6
MFCD11035888
2-oxa-7-azaspiro 3.5 nonane oxalate 2:1, 2-oxa-7-azaspiro 3.5 nonane hemioxalate, bis 2-oxa-7-azaspiro 3.5 nonane ; oxalic acid, 2-oxa-7-azaspiro 3.5 nonan hemioxalate, 2-oxa-7-azaspiro 3,5 nonane hemioxalate, bis 2-oxa-7-azaspiro 3.5 nonane oxalate, 2-oxa-7-azaspiro 3.5 nonane hemi-oxalate
2-oxa-7-azaspiro[3.5]nonane;oxalic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only