missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(p-Tolyl)ethanol 98.0+%, TCI America™
Supplier: TCI America M229825G
Specifications
2-(p-Tolyl)ethanol | |
Yellow | |
C9H12O | |
25 g | |
DAVFJRVIVZOKKS-UHFFFAOYSA-N | |
2-(4-methylphenyl)ethanol | |
95294 | |
≥98.0% (GC) |
699-02-5 | |
115°C | |
MFCD00002904 | |
2-4-methylphenyl ethanol, 2-p-tolyl ethanol, 4-methylphenethyl alcohol, benzeneethanol, 4-methyl, 2-p-tolylethanol, p-methylphenethyl alcohol, 4-methylbenzeneethanol, 2-4-methylphenyl ethan-1-ol, p-methylphenylethyl alcohol, 2-p-methylphenyl ethanol | |
CC1=CC=C(C=C1)CCO | |
136.194 | |
136.19 | |
Liquid |
Chemical Identifiers
699-02-5 | |
136.194 | |
DAVFJRVIVZOKKS-UHFFFAOYSA-N | |
95294 | |
CC1=CC=C(C=C1)CCO |
C9H12O | |
MFCD00002904 | |
2-4-methylphenyl ethanol, 2-p-tolyl ethanol, 4-methylphenethyl alcohol, benzeneethanol, 4-methyl, 2-p-tolylethanol, p-methylphenethyl alcohol, 4-methylbenzeneethanol, 2-4-methylphenyl ethan-1-ol, p-methylphenylethyl alcohol, 2-p-methylphenyl ethanol | |
2-(4-methylphenyl)ethanol |
Safety and Handling
EINECSNumber : (3)-1068
TSCA : Yes