missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(p-Tolyl)indole 98.0+%, TCI America™
$320.62 - $967.44
Chemical Identifiers
| CAS | 55577-25-8 |
|---|---|
| Molecular Formula | C15H13N |
| Molecular Weight (g/mol) | 207.28 |
| MDL Number | MFCD00087262 |
| InChI Key | VPXGIHGJJJBJFP-UHFFFAOYSA-N |
| Synonym | 2-(4-Methylphenyl)indole |
| PubChem CID | 292599 |
| IUPAC Name | 2-(4-methylphenyl)-1H-indole |
| SMILES | CC1=CC=C(C=C1)C1=CC2=CC=CC=C2N1 |
Chemical Identifiers
| 55577-25-8 | |
| 207.28 | |
| VPXGIHGJJJBJFP-UHFFFAOYSA-N | |
| 292599 | |
| CC1=CC=C(C=C1)C1=CC2=CC=CC=C2N1 |
| C15H13N | |
| MFCD00087262 | |
| 2-(4-Methylphenyl)indole | |
| 2-(4-methylphenyl)-1H-indole |
Specifications
| 55577-25-8 | |
| White-Yellow | |
| MFCD00087262 | |
| 2-(4-Methylphenyl)indole | |
| CC1=CC=C(C=C1)C1=CC2=CC=CC=C2N1 | |
| 207.28 | |
| 207.28 | |
| Crystalline Powder |
| 222°C | |
| C15H13N | |
| 1 g | |
| VPXGIHGJJJBJFP-UHFFFAOYSA-N | |
| 2-(4-methylphenyl)-1H-indole | |
| 292599 | |
| ≥98.0% (GC) | |
| 2-(p-Tolyl)indole |
Safety and Handling
TSCA : No