Learn More
2-Pentene, cis + trans, 98%, Thermo Scientific Chemicals
$173.66 - $507.67
Chemical Identifiers
CAS | 109-68-2 |
---|---|
Molecular Formula | C5H10 |
Molecular Weight (g/mol) | 70.14 |
MDL Number | MFCD00009384 |
InChI Key | QMMOXUPEWRXHJS-HYXAFXHYSA-N |
Synonym | trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e |
PubChem CID | 5326161 |
SMILES | CC\C=C/C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1773614
|
Thermo Scientific Chemicals
A1773614 |
25 g |
N/A
|
|
||||||
AAA1773622
|
Thermo Scientific Chemicals
A1773622 |
100 g |
N/A
|
|
||||||
Description
2-Pentene (mixture of cis and trans) is used in the synthesis of polycyclopentene by ring-opening metathesis polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Pentene (mixture of cis and trans) is used in the synthesis of polycyclopentene by ring-opening metathesis polymerization.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
Chemical Identifiers
109-68-2 | |
70.14 | |
QMMOXUPEWRXHJS-HYXAFXHYSA-N | |
5326161 |
C5H10 | |
MFCD00009384 | |
trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e | |
CC\C=C/C |
Specifications
109-68-2 | |
36°C to 38°C | |
Petroleum-like | |
1.38 | |
MFCD00009384 | |
UN3295 | |
14,7123 | |
Not miscible or difficult to mix in water. | |
CC\C=C/C | |
70.14 | |
70.14 | |
2-Pentene, cis + trans |
0.65 | |
−45°C (−49°F) | |
C5H10 | |
CH3CH2CH=CHCH3 | |
25 g | |
969151 | |
trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e | |
QMMOXUPEWRXHJS-HYXAFXHYSA-N | |
(2Z)-pent-2-ene | |
5326161 | |
98% |
Safety and Handling
GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
H225-H304
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 203-695-0
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only