missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Phenoxyethanol, 99%, Thermo Scientific Chemicals

2-Phenoxyethanol, 99%, C8H10O2, CAS Number-122-99-6, ethylene glycol monophenyl ether, ethanol, 2-phenoxy, 1-hydroxy-2-phenoxyethane, phenoxyethanol, phenoxyethyl alcohol, phenoxethol, phenoxytol, phenoxetol, phenyl cellosolve, ethylene glycol phenyl ether, 25L, 98.5% min. (GC), 06,146, 237.0 deg.C

Supplier:  Thermo Scientific Chemicals 130240250

 View more versions of this product

Catalog No. AC130240250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Phenoxyethanol
98.5
11.0°C to 13.0°C
1.1000g/mL
130°C
98.5% min. (GC)
C8H10O2
C6H5OCH2CH2OH
25 L
1.1
phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane
QCDWFXQBSFUVSP-UHFFFAOYSA-N
2-phenoxyethanol
31236
CHEBI:64275
99%
122-99-6
100.0
Colorless
237.0°C
Authentic
Metal drum
1.5360 to 1.5400
MFCD00002857
06,146
15,7369
Solubility in water: 30g/L (20°C). Other solubilities: freely soluble in alcohol and ether,miscible with acetone and ethyl acetate
C1=CC=C(C=C1)OCCO
138.17
29 mPa.s (20°C)
138.17
Liquid

Chemical Identifiers

122-99-6
138.17
QCDWFXQBSFUVSP-UHFFFAOYSA-N
31236
2-phenoxyethanol
C8H10O2
MFCD00002857
phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane
CHEBI:64275
C1=CC=C(C=C1)OCCO
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-589-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only