missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Phenyl-1,3-thiazole-4-carbaldehyde, ≥90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC18304DA

 View more versions of this product

Catalog No. CC18304DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2-Phenyl-1, 3-thiazole-4-carbaldehyde
94.5
95%
C10H7NOS
1g
OLLKCCGWRITPOV-UHFFFAOYSA-N
2-phenyl-1,3-thiazole-4-carbaldehyde
736524
95%
20949-81-9
100.0
Amber Glass Bottle
MFCD02681934
2-phenylthiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, 2-phenyl, 2-phenyl-4-formylthiazole, 4-formyl-2-phenyl-1,3-thiazole, 4-thiazolecarboxaldehyde,2-phenyl, 2-phenyl-thiazole-4-carbaldehyde, 2-phenyl-4-thiazolecarboxaldehyde, 4-formyl-1,3-thiazol-2-yl benzene, 2-phenyl-1,3-thiazole-4-carboxaldehyde
C1=CC=C(C=C1)C2=NC(=CS2)C=O
189.232
189.24

Chemical Identifiers

20949-81-9
189.232
OLLKCCGWRITPOV-UHFFFAOYSA-N
736524
C1=CC=C(C=C1)C2=NC(=CS2)C=O
C10H7NOS
MFCD02681934
2-phenylthiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, 2-phenyl, 2-phenyl-4-formylthiazole, 4-formyl-2-phenyl-1,3-thiazole, 4-thiazolecarboxaldehyde,2-phenyl, 2-phenyl-thiazole-4-carbaldehyde, 2-phenyl-4-thiazolecarboxaldehyde, 4-formyl-1,3-thiazol-2-yl benzene, 2-phenyl-1,3-thiazole-4-carboxaldehyde
2-phenyl-1,3-thiazole-4-carbaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions