missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+/-)-2-Phenyl-1-propanol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2347330

 View more versions of this product

Catalog No. AAB2347330


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(+/-)-2-Phenyl-1-propanol
-37°C
220°C
C9H12O
MFCD00004736
1906760
RNDNSYIPLPAXAZ-UHFFFAOYSA-N
2-phenylpropan-1-ol
14295
97%
1123-85-9
0.975
108°C (226°F)
1.5262
250 g
2-phenyl-1-propanol, hydratropic alcohol, hydratropyl alcohol, 2-phenylpropanol-1, beta-methylphenethyl alcohol, 2-phenylpropyl alcohol, 1-propanol, 2-phenyl, 1-hydroxy-2-phenylpropane, 2-phenylpropanol, benzeneethanol, .beta.-methyl
CC(CO)C1=CC=CC=C1
136.194
136.19

Chemical Identifiers

1123-85-9
136.194
RNDNSYIPLPAXAZ-UHFFFAOYSA-N
14295
CC(CO)C1=CC=CC=C1
C9H12O
MFCD00004736
2-phenyl-1-propanol, hydratropic alcohol, hydratropyl alcohol, 2-phenylpropanol-1, beta-methylphenethyl alcohol, 2-phenylpropyl alcohol, 1-propanol, 2-phenyl, 1-hydroxy-2-phenylpropane, 2-phenylpropanol, benzeneethanol, .beta.-methyl
2-phenylpropan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 214-379-7

RTECSNumber : SG8590000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only