missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Phenyl-2-imidazoline, 98%, Thermo Scientific Chemicals

$34.34 - $69.18

Chemical Identifiers

CAS 936-49-2
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00005180
InChI Key BKCCAYLNRIRKDJ-UHFFFAOYSA-N
Synonym 2-phenyl-2-imidazoline, 2-phenylimidazoline, 1h-imidazole, 4,5-dihydro-2-phenyl, 2-imidazoline, 2-phenyl, 4,5-dihydro-2-phenyl-1h-imidazole, 2-phenyl imidazoline, 2-phenyl-2-imidazolin, acmc-209rmt, labotest-bb lt00000058, 5-23-06-00425 beilstein handbook reference
PubChem CID 13639
IUPAC Name 2-phenyl-4,5-dihydro-1H-imidazole
SMILES C1CN=C(N1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5909509
View Documents
Thermo Scientific Chemicals
H5909509
10 g
Each for $34.34
 
AAH5909518
View Documents
Thermo Scientific Chemicals
H5909518
50 g
Each for $69.18
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

936-49-2
146.19
BKCCAYLNRIRKDJ-UHFFFAOYSA-N
13639
C1CN=C(N1)C1=CC=CC=C1
C9H10N2
MFCD00005180
2-phenyl-2-imidazoline, 2-phenylimidazoline, 1h-imidazole, 4,5-dihydro-2-phenyl, 2-imidazoline, 2-phenyl, 4,5-dihydro-2-phenyl-1h-imidazole, 2-phenyl imidazoline, 2-phenyl-2-imidazolin, acmc-209rmt, labotest-bb lt00000058, 5-23-06-00425 beilstein handbook reference
2-phenyl-4,5-dihydro-1H-imidazole

Specifications

936-49-2
Brown
C9H10N2
10 g
2-phenyl-2-imidazoline, 2-phenylimidazoline, 1h-imidazole, 4,5-dihydro-2-phenyl, 2-imidazoline, 2-phenyl, 4,5-dihydro-2-phenyl-1h-imidazole, 2-phenyl imidazoline, 2-phenyl-2-imidazolin, acmc-209rmt, labotest-bb lt00000058, 5-23-06-00425 beilstein handbook reference
C1CN=C(N1)C1=CC=CC=C1
146.19
146.19
Powder
99°C to 104°C
201°C (394°F)
MFCD00005180
119250
BKCCAYLNRIRKDJ-UHFFFAOYSA-N
2-phenyl-4,5-dihydro-1H-imidazole
13639
98%
2-Phenyl-2-imidazoline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 213-313-4

RTECSNumber : NJ4395500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only