missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Phenyl-2-pentenal, (E)+(Z), 90+%, Thermo Scientific Chemicals

$98.73 - $348.29

Chemical Identifiers

CAS 3491-63-2
Molecular Formula C11H12O
Molecular Weight (g/mol) 160.216
MDL Number MFCD00051835
InChI Key YPAJRUMMODCONM-IZZDOVSWSA-N
Synonym 2-phenyl-2-pentenal, 2z-2-phenylpent-2-enal
PubChem CID 15310469
IUPAC Name (Z)-2-phenylpent-2-enal
SMILES CCC=C(C=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1363506
View Documents
Thermo Scientific Chemicals
A1363506
5 g
Each for $98.73
 
AAA1363514
View Documents
Thermo Scientific Chemicals
A1363514
25 g
Each for $348.29
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3491-63-2
160.216
YPAJRUMMODCONM-IZZDOVSWSA-N
15310469
CCC=C(C=O)C1=CC=CC=C1
C11H12O
MFCD00051835
2-phenyl-2-pentenal, 2z-2-phenylpent-2-enal
(Z)-2-phenylpent-2-enal

Specifications

3491-63-2
124°C to 126°C (14 mmHg)
C11H12O
MFCD00051835
1859032
2-phenyl-2-pentenal, 2z-2-phenylpent-2-enal
CCC=C(C=O)C1=CC=CC=C1
160.216
160.22
2-Phenyl-2-pentenal, (E)+(Z)
1.005
>100°C (212°F)
1.551
5 g
Air sensitive
YPAJRUMMODCONM-IZZDOVSWSA-N
(Z)-2-phenylpent-2-enal
15310469
≥90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only