missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Phenyl-2-propanol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 130830250

 View more versions of this product

Catalog No. AC130830250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Phenyl-2-propanol
98.5
32°C to 34°C
0.9730g/mL
88°C
98.5% min. (GC)
C9H12O
C6H5C(CH3)2OH
25 g
0.973
Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether,benzene
CC(C)(O)C1=CC=CC=C1
136.19
136.19
Liquid After Melting
617-94-7
100.0
Colorless to Yellow
88°C to 90°C (11.0 mmHg)
Authentic
Glass bottle
1.5190 to 1.522
MFCD00004456
06, 506
2-phenyl-2-propanol, dimethylphenylcarbinol, 1-hydroxycumene, dimethylphenylmethanol, alpha-cumyl alcohol, phenyldimethylcarbinol, 2-propanol, 2-phenyl, 2-phenylisopropanol, alpha,alpha-dimethylbenzyl alcohol, 2-phenyl-propan-2-ol
BDCFWIDZNLCTMF-UHFFFAOYSA-N
2-phenylpropan-2-ol
12053
99%

Chemical Identifiers

617-94-7
136.19
BDCFWIDZNLCTMF-UHFFFAOYSA-N
12053
CC(C)(O)C1=CC=CC=C1
C9H12O
MFCD00004456
2-phenyl-2-propanol, dimethylphenylcarbinol, 1-hydroxycumene, dimethylphenylmethanol, alpha-cumyl alcohol, phenyldimethylcarbinol, 2-propanol, 2-phenyl, 2-phenylisopropanol, alpha,alpha-dimethylbenzyl alcohol, 2-phenyl-propan-2-ol
2-phenylpropan-2-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

GHS Signal Word: Warning

EINECSNumber : 210-539-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only