missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Phenylacetamide 98.0+%, TCI America™

Supplier:  TCI America P0120500G

 View more versions of this product

Catalog No. P0120500G


Explore available promotions

Description

Description

Specifications

Specifications

2-Phenylacetamide
156°C
C8H9NO
500 g
LSBDFXRDZJMBSC-UHFFFAOYSA-N
2-phenylacetamide
7680
135.17
Crystalline Powder
103-81-1
White
MFCD00059193
benzeneacetamide, phenylacetamide, alpha-toluamide, alpha-phenylacetamide, phenylacetic acid amide, acetamide, 2-phenyl, phenyl-beta-acetylamine, alpha-toluimidic acid, 2-phenyl-acetamide, phenyl acetamide
NC(=O)CC1=CC=CC=C1
135.17
CHEBI:16562
≥98.0% (N)

Chemical Identifiers

103-81-1
135.17
LSBDFXRDZJMBSC-UHFFFAOYSA-N
7680
2-phenylacetamide
C8H9NO
MFCD00059193
benzeneacetamide, phenylacetamide, alpha-toluamide, alpha-phenylacetamide, phenylacetic acid amide, acetamide, 2-phenyl, phenyl-beta-acetylamine, alpha-toluimidic acid, 2-phenyl-acetamide, phenyl acetamide
CHEBI:16562
NC(=O)CC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1200

RTECSNumber : AC7705000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions