Learn More
2-Phenylethanol, 98+%, Thermo Scientific Chemicals
$77.59 - $687.93
Chemical Identifiers
CAS | 60-12-8 |
---|---|
Molecular Formula | C8H10O |
Molecular Weight (g/mol) | 122.17 |
MDL Number | MFCD00002886 |
InChI Key | WRMNZCZEMHIOCP-UHFFFAOYSA-N |
Synonym | phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol |
PubChem CID | 6054 |
ChEBI | CHEBI:49000 |
SMILES | OCCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1524130
|
Thermo Scientific Chemicals
A1524130 |
250 g |
Each for $77.59
|
|
|||||
AAA152410B
|
Thermo Scientific Chemicals
A152410B |
1000 g |
Each for $184.96
|
|
|||||
AAA152410I
|
Thermo Scientific Chemicals
A152410I |
5000 g |
Each for $687.93
|
|
|||||
Description
2-Phenylethanol is used also as an aromatic essence and preservative in pharmaceutics and perfumery. It is also used as odor agents, air care products, cleaning and furnishing care products, laundry and dishwashing products, personal care products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Phenylethanol is used also as an aromatic essence and preservative in pharmaceutics and perfumery. It is also used as odor agents, air care products, cleaning and furnishing care products, laundry and dishwashing products, personal care products.
Solubility
Miscible with chloroform.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
60-12-8 | |
122.17 | |
WRMNZCZEMHIOCP-UHFFFAOYSA-N | |
6054 | |
OCCC1=CC=CC=C1 |
C8H10O | |
MFCD00002886 | |
phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol | |
CHEBI:49000 |
Specifications
60-12-8 | |
1.021 | |
101°C (214°F) | |
C8H10O | |
C6H5(CH2)2OH | |
250 g | |
1905732 | |
phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol | |
WRMNZCZEMHIOCP-UHFFFAOYSA-N | |
2-phenylethanol | |
6054 | |
122.17 | |
Liquid |
°C to 27°C | |
219°C | |
≥98% | |
1.533 | |
MFCD00002886 | |
UN2810 | |
14,7224 | |
Miscible with chloroform. | |
OCCC1=CC=CC=C1 | |
122.17 | |
CHEBI:49000 | |
≥98% | |
2-Phenylethanol |
Safety and Handling
GHS H Statement
H311-H302-H315-H319
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 200-456-2
RTECSNumber : SG7175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only