missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Phenylethyl acetate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2123830

 View more versions of this product

Catalog No. AAB2123830


Explore available promotions

Description

Description

Phenylethyl acetate mantains the typical rose notes of Phenylethanol with a more intense honey profile which is perfect for fruit and alcoholic drink applications. It is used as a pharmaceutical intermediate and also it holds application in gas chromatography and liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylethyl acetate mantains the typical rose notes of Phenylethanol with a more intense honey profile which is perfect for fruit and alcoholic drink applications. It is used as a pharmaceutical intermediate and also it holds application in gas chromatography and liquid chromatography.

Solubility
It is insoluble in paraffin oil, water. Soluble in alcohol, dipropylene glycol, fixed oils, kerosene, propylene glycol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Protected from heat and light.
Specifications

Specifications

2-Phenylethyl acetate
-31°C
231°C to 233°C
Fruit-like
1.499
250 g
phenethyl acetate, 2-phenethyl acetate, acetic acid, 2-phenylethyl ester, benzylcarbinyl acetate, beta-phenylethyl acetate, acetic acid, phenethyl ester, phenethyl alcohol, acetate, phenylethyl acetate, acetic acid phenethyl ester, ethanol, 2-phenyl-, acetate
MDHYEMXUFSJLGV-UHFFFAOYSA-N
2-phenylethyl acetate
7654
164.21
103-45-7
1.029
107°C (225°F)
C10H12O2
MFCD00008720
638179
It is insoluble in paraffin oil,water. Soluble in alcohol,dipropylene glycol,fixed oils,kerosene,propylene glycol.
CC(=O)OCCC1=CC=CC=C1
164.20
CHEBI:31988
98%

Chemical Identifiers

103-45-7
164.20
MDHYEMXUFSJLGV-UHFFFAOYSA-N
7654
2-phenylethyl acetate
C10H12O2
MFCD00008720
phenethyl acetate, 2-phenethyl acetate, acetic acid, 2-phenylethyl ester, benzylcarbinyl acetate, beta-phenylethyl acetate, acetic acid, phenethyl ester, phenethyl alcohol, acetate, phenylethyl acetate, acetic acid phenethyl ester, ethanol, 2-phenyl-, acetate
CHEBI:31988
CC(=O)OCCC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-113-5

RTECSNumber : AJ2220000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only