Learn More
2-Phenylethyl isothiocyanate, 98%
CAS: 2257-09-2 | C9H9NS | 163.24 g/mol
$315.99 - $315.99
Chemical Identifiers
| CAS | 2257-09-2 |
|---|---|
| Molecular Formula | C9H9NS |
| Molecular Weight (g/mol) | 163.24 |
| MDL Number | MFCD00004821 |
| InChI Key | IZJDOKYDEWTZSO-UHFFFAOYSA-N |
| Synonym | 2-phenylethyl isothiocyanate, phenethyl isothiocyanate, phenylethyl isothiocyanate, 2-isothiocyanatoethyl benzene, phenethyl mustard oil, phenylaethylsenfoel, benzene, 2-isothiocyanatoethyl, peitc, phenylethyl mustard oil, 2-phenylethylisothiocyanate |
| PubChem CID | 16741 |
| ChEBI | CHEBI:351346 |
| IUPAC Name | 2-isothiocyanatoethylbenzene |
| SMILES | C1=CC=C(C=C1)CCN=C=S |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1512314
|
Thermo Scientific Chemicals
A1512314 |
25 g |
N/A
|
|
|||||
Description
Phenethyl isothiocyanate is an NNK-induced lung tumorigenesis inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenethyl isothiocyanate is an NNK-induced lung tumorigenesis inhibitor.
Solubility
Soluble in chloroform, ethyl acetate, and dichoromethane. Reacts with water.
Notes
Moisture Sensitive, store away from moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agent. Store under dry inert gas.
Chemical Identifiers
| 2257-09-2 | |
| 163.24 | |
| IZJDOKYDEWTZSO-UHFFFAOYSA-N | |
| 16741 | |
| 2-isothiocyanatoethylbenzene |
| C9H9NS | |
| MFCD00004821 | |
| 2-phenylethyl isothiocyanate, phenethyl isothiocyanate, phenylethyl isothiocyanate, 2-isothiocyanatoethyl benzene, phenethyl mustard oil, phenylaethylsenfoel, benzene, 2-isothiocyanatoethyl, peitc, phenylethyl mustard oil, 2-phenylethylisothiocyanate | |
| CHEBI:351346 | |
| C1=CC=C(C=C1)CCN=C=S |
Specifications
| 2257-09-2 | |
| 139°C to 140°C (11 mmHg) | |
| Pungent | |
| 1.5888 | |
| 25 g | |
| 2084162 | |
| 2-phenylethyl isothiocyanate, phenethyl isothiocyanate, phenylethyl isothiocyanate, 2-isothiocyanatoethyl benzene, phenethyl mustard oil, phenylaethylsenfoel, benzene, 2-isothiocyanatoethyl, peitc, phenylethyl mustard oil, 2-phenylethylisothiocyanate | |
| IZJDOKYDEWTZSO-UHFFFAOYSA-N | |
| 2-isothiocyanatoethylbenzene | |
| 16741 | |
| 163.24 | |
| 2-Phenylethyl isothiocyanate |
| 1.09 | |
| >110°C (230°F) | |
| C9H9NS | |
| MFCD00004821 | |
| UN2922 | |
| Moisture sensitive | |
| Soluble in chloroform,ethyl acetate,and dichoromethane. Reacts with water. | |
| C1=CC=C(C=C1)CCN=C=S | |
| 163.24 | |
| CHEBI:351346 | |
| 98% |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P501c
H302+H312+H332-H315-H319-H334-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 218-855-5
RTECSNumber : NX9115000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only