missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Phenylethylamine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A13402AD

 View more versions of this product

Catalog No. AAA13402AD


Explore available promotions

Description

Description

Phenethylamine is used in manufacturing anti-depression agents and anti diabetic drugs. It is also used as the goodds for drug jiangtangling intermediate, also used for other organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenethylamine is used in manufacturing anti-depression agents and anti diabetic drugs. It is also used as the goodds for drug jiangtangling intermediate, also used for other organic synthesis.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air sensitive. Store under inert gas. Keep away from acids, Acid chlorides, Acid anhydrides, Strong oxidizing agents, Carbon dioxide (CO2).
Specifications

Specifications

2-Phenylethylamine
-60°C
11.5
81°C (178°F)
C8H11N
C6H5(CH2)2NH2
50 mL
507488
14,7225
It is soluble in water.
C1=CC=C(C=C1)CCN
121.183
CHEBI:18397
99%
64-04-0
0.964
200°C to 202°C
Amine-like
1.5335
MFCD00008184
UN2922
Air Sensitive
phenethylamine, 2-phenylethylamine, benzeneethanamine, phenylethylamine, 2-phenethylamine, beta-phenylethylamine, beta-phenethylamine, 1-amino-2-phenylethane, 2-aminoethyl benzene, beta-aminoethylbenzene
BHHGXPLMPWCGHP-UHFFFAOYSA-N
2-phenylethanamine
1001
121.18

Chemical Identifiers

64-04-0
121.183
BHHGXPLMPWCGHP-UHFFFAOYSA-N
1001
2-phenylethanamine
C8H11N
MFCD00008184
phenethylamine, 2-phenylethylamine, benzeneethanamine, phenylethylamine, 2-phenethylamine, beta-phenylethylamine, beta-phenethylamine, 1-amino-2-phenylethane, 2-aminoethyl benzene, beta-aminoethylbenzene
CHEBI:18397
C1=CC=C(C=C1)CCN
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H301-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: I

EINECSNumber : 200-574-4

RTECSNumber : SG8750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only