missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Phenylethylamine Hydrobromide 98.0+%, TCI America™

Supplier:  TCI America P23885G

 View more versions of this product

Catalog No. P23885G


Explore available promotions

Description

Description

Specifications

Specifications

2-Phenylethylamine Hydrobromide
259°C
C8H12BrN
Phenethylamine Hydrobromide, 2-Phenylethylammonium Bromide
C1=CC=C(C=C1)CCN.Br
202.095
202.10
Crystalline Powder
53916-94-2
White
5 g
IRAGENYJMTVCCV-UHFFFAOYSA-N
2-phenylethanamine;hydrobromide
70441016
≥98.0% (HPLC,T)

Chemical Identifiers

53916-94-2
202.095
Phenethylamine Hydrobromide, 2-Phenylethylammonium Bromide
2-phenylethanamine;hydrobromide
C8H12BrN
IRAGENYJMTVCCV-UHFFFAOYSA-N
70441016
C1=CC=C(C=C1)CCN.Br
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions